Skip to Content
Merck
All Photos(1)

Key Documents

Z711004

Media bottles, wide mouth, Lab 45, accessory

polypropylene screw cap for 45mm

Synonym(s):

240726

Sign Into View Organizational & Contract Pricing

Select a Size

12 EA
€46.60

€46.60


Check Cart for Availability


Select a Size

Change View
12 EA
€46.60

About This Item

UNSPSC Code:
41121800
NACRES:
NB.17

€46.60


Check Cart for Availability

material

polypropylene screw cap (for 45mm)

manufacturer/tradename

WHEATON®

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • For high temperature applications, use the PBT (polybutylene terephthalate) screw caps and the ETFE (ethylene tetrafluoroethylene) pour lips which can withstand temperatures up to 200ºC up to 2 hours
  • Autoclavable

Legal Information

WHEATON is a registered trademark of Wheaton Industries, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service