Key Documents
Z710202
Nichipet Eco pipette
volume 1000-5000 μL
Select a Size
1 EA
€614.00
€614.00
Check Cart for Availability
Select a Size
Change View
1 EA
€614.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
€614.00
Check Cart for Availability
description
With glass tip
feature
electronic: no
single-channel
packaging
pack of 1 ea
manufacturer/tradename
Nichiryo 00-NPC-5000
volume measuring range
1000-5000 μL
volume
5000 μL
General description
Providing the accuracy and precision of a micropipette with the compatibility of glass. Glass tips allow pipetting of substances not compatibile with plastic.
  • Glass tip enables totally bias free liquid handling for endocrine disruptors analysis
  • 4 sizes cover a wide volume range (20-10000 μL)
  • Nozzle with double O-rings enables easy tip fitting/ejection
  • Reliable sealing nozzle is PTFE and piston is ceramic
Choose from one of the most recent versions:
Certificates of Analysis (COA)