Skip to Content
Merck

Skip To

Z709166

ART® self-sealing barrier pipette tips

ART GEL, volume range 0.5-100 μL, sterile

Synonym(s):

pipet tips, pipette tips, MBP 2155, dispensing tip, liquid transfer tip

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 pkg
Check Cart for Availability
€ 218,00
4 pkg
Check Cart for Availability
€ 785,00
€ 667,25

About This Item

NACRES:
NB.01
UNSPSC Code:
41121601
Volume:
100 μL
Sterility:
sterile
Feature:
DNase free, RNase free, barrier
Packaging:
pack of 960 ea
Manufacturer/tradename:
Molecular BioProducts 2155

€ 218,00


Check Cart for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

colorless , colorless tip

sterility

sterile

feature

DNase free, RNase free, barrier

packaging

pack of 960 ea

manufacturer/tradename

Molecular BioProducts 2155

technique(s)

PCR: suitable

maximum volume

100 μL

volume range

0.5-100 μL

volume

100 μL

compatibility

for use with

  • Biohit Proline-Electronic 10-100 μl
  • Biohit Proline-Variable 50-200 μl
  • Eppendorf 4810 10-100 μl
  • Eppendorf Research Pro 5-100 μl
  • Eppendorf Research/Reference 10-100 μl
  • Eppendorf Response 4850 5-100 μl
  • Finnpipette 4027/4500 Digital 5-40 μl
  • Mettler-Toledo VoluMate 10-100 μl
  • Pipetman P-100 10-100 μl
  • Rainin EDP 2 10-100ul/20-250 μl
  • Rainin EDP Plus 10-100ul/20-250 μl
  • Rainin Pipet Plus 10-100ul/20-200 μl
  • Rainin Pipetman 10-100ul/20-200 μl
  • Transferpette (Limited Draw) 20-200 μl

General description

Load Agarose and polyacrylamide gels up to 100 μl

Packaging

960EA = 10 trays of 96 tips (1 pack)
4800EA = 5 packs of 960 tips (50 trays of 96 tips)

800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA

Other Notes

Browse all pipette tips

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Z569771Z717177A2598
description

ART GEL, volume range 0.5-100 μL, sterile

description

ART 10 Reach, volume range 0.1-10 μL, sterile

description

ART 20, volume range 0.5-20 μL, sterile

description

ART 10, volume range 0.1-10 μL, sterile

volume

100 μL

volume

0.1—10 μL

volume

0.5—20 μL

volume

0.1—10 μL

feature

RNase free, DNase free, barrier

feature

barrier, RNase free, DNase free

feature

RNase free, barrier, DNase free

feature

barrier, RNase free, DNase free

sterility

sterile

sterility

sterile

sterility

sterile

sterility

sterile

manufacturer/tradename

Molecular BioProducts 2155

manufacturer/tradename

Molecular BioProducts 2140

manufacturer/tradename

Molecular BioProducts 2149

manufacturer/tradename

Molecular BioProducts 2139

material

colorless , colorless tip

material

colorless

material

colorless tip

material

colorless

packaging

pack of 960 ea

packaging

pack of 960 ea

packaging

pack of 960 ea (10 racks of 96 tips)

packaging

case of 5 packs, pack of 960 ea (racked)


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library