Skip to Content
Merck
All Photos(1)

Key Documents

Z672548

Advanced PAP Pen

Advanced PAP Pen
1 of 3 reviewers received a sample product or took part in a promotion

2 mm tip width

Synonym(s):

hydrophobic barrier pen, immunostaining wax pen, liquid blocker pap pen, water repellant barrier pen

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€127.00

€127.00


Check Cart for Availability


Select a Size

Change View
1 EA
€127.00

About This Item

UNSPSC Code:
41181600
NACRES:
MA.01

€127.00


Check Cart for Availability

packaging

pack of 1 ea

manufacturer/tradename

Ted Pella 22311

Looking for similar products? Visit Product Comparison Guide

Application

Use pen to draw a hydrophobic circle around slide-mounted tissue. Prevents the waste of valuable reagents by keeping liquid pooled in a small droplet. Insoluble in ethanol, acetone and water, but will wash off with xylene.

For use with paraffin-mounted sections or slides treated with albumin or poly-lysine.

Quantity

Each pen can make more than 600 circles.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Ozone 1 - Repr. 1B - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Liver,Central nervous system, Respiratory system

WGK

WGK 3

Flash Point(F)

159.8 °F

Flash Point(C)

71 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

EU REACH Annex XVII (Restriction List)

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gopinath Krishnamoorthy et al.
Disease models & mechanisms, 13(3) (2020-02-09)
Lactate dehydrogenase A (LDHA) mediates interconversion of pyruvate and lactate, and increased lactate turnover is exhibited by malignant and infected immune cells. Hypoxic lung granuloma in Mycobacterium tuberculosis-infected animals present elevated levels of Ldha and lactate. Such alterations in the

Questions

1–7 of 7 Questions  
  1. Will it hold in high temperatures? what is the highest temperature it can still be properly used in?

    1 answer
    1. This pen is stable at temperatures up to 120°C

      Helpful?

  2. Is there a hydrophobic pen available for immunofluorescence?

    1 answer
    1. Yes, there are hydrophobic pens available for immunofluorescence. The catalog numbers for the available pens are Z672548 and Z377821, and they are named Advanced PAP pen.

      Helpful?

  3. Will mounting a coverslip be a problem with the pap pen if my assay does not include a xylene step that removes this liquid barrier?

    1 answer
    1. This product is suitable for use with coverslip mounts so long as xylene is avoided.

      Helpful?

  4. Is this pen suitable to contain a drop of 3% H2O2 in methanol solution? Could the circle be erased by methanol?

    1 answer
    1. Yes, this product is resistant to most organic solvents and would not be affected by exposure to methanol. However, the barrier may be dissolved by xylene.

      Helpful?

  5. Hi, I have slide with 4x5 tissue spots of 5 mm size each. I like to draw hydrophobic barrier line to protect each tissue spots by drawing a column or row or grid. Therefore, i would require a fine tip Pap pen. Is 2 mm tip good or 1 mm will be better.

    1 answer
    1. The advanced PAP Pen is commonly used to create hydrophobic barriers on slides for various laboratory applications. It is often used to outline areas and draw boundaries to prevent mixing of reagents or solutions. Regarding the tip size, it depends on the specific requirements and preferences of the end-user. Both 2 mm and 1 mm tips can be suitable for creating hydrophobic barriers of the desired size on your slides. The choice between the two will depend on factors such as the precision needed by the end-user, the desired thickness or width of the barrier, and the personal experience and comfort with using different tip sizes of the end-user's.

      If a finer and more precise line is required, a 1 mm tip may be preferable. However, if a slightly wider or more visible barrier line is needed, the 2 mm tip can be suitable.

      Helpful?

  6. How much volume does the pen contain? I couldn't find it in the specification sheet. Thanks,

    1 answer
    1. This pen contains approximately 7.5 mL of liquid. The volume is sufficient to place more than 600 circles.

      Helpful?

  7. Does this pen marking autofluorese?

    1 answer
    1. This pen is suitable for use with fluorescent antibodies. It is not specifically tested for autofluorescence; no reports or complaints of this occurring have been filed. This PAP Pen Liquid Blocker, is a hydrophobic slide marker for staining procedures, creates a visible hydrophobic barrier that provides proper surface tension to hold reagents within a targeted area when applied around tissue sections or smears on a microscopic slide. This surface tension ensures the amount of reagent needed for sufficient reaction is reduced and retained for complete tissue section coverage. Multiple specimens can be separated by drawn circles or lines applied to the same slide.

      PAP Pen Liquid Blockers contain a unique formulation that is water repellent, insoluble in alcohol and acetone, soluble in xylene and temperature resistant up to 120°C.

      Helpful?

Reviews

1 of 3 reviewers received a sample product or took part in a promotion

Active Filters

  1. Portugal
    • Review 1
    • Votes 0
    4 out of 5 stars.

    Useful

    It is so useful in IHC because it allows you to use less ab

    Helpful?

  2. 2 Ratings-Only Reviews

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service