Key Documents
Z659401
Bandelin SONOPULS ultrasonic homogenizer accessories
Indirect processing Cup horn BB 6 for use with HD 2200 homogenizer
Synonym(s):
sonication vessel
Select a Size
1 EA
€1,750.00
€1,750.00
Check Cart for Availability
Select a Size
Change View
1 EA
€1,750.00
About This Item
UNSPSC Code:
41101520
NACRES:
NB.74
€1,750.00
Check Cart for Availability
packaging
pack of 1 ea
manufacturer/tradename
Bandelin 3605
availability
available only in EU
General description
BB6 for indirect irradiation with high energy. No contamination from cavitation erosion. No aerosoling with pathogenic or hazardous materials. Temperature regulation with circulating liquid coolants.
Choose from one of the most recent versions:
Certificates of Analysis (COA)