Z640522
Eppendorf® epT.I.P.S. Reloads, PCR Clean
volume 50-1000 μL
Synonym(s):
pipetting tips
About This Item
Eppendorf® 022491750
Recommended Products
sterility
non-sterile
packaging
pack of 960 (10 trays of 96 tips)
manufacturer/tradename
Eppendorf® 0030 073.843
Eppendorf® 022491750
volume measuring range
50-1000 μL
technique(s)
PCR: suitable
L
71 mm
maximum volume
1 mL
volume
50—1000 μL
Looking for similar products? Visit Product Comparison Guide
General description
Trays of tips which may be autoclaved in their packaging and stored clean for later use. Tips may be used straight from the reload tray, or loaded into ep T.I.P.S box (purchase separately). PCR clean, free of human DNA, DNase, RNase and PCR inhibitors, batch related certificates available on the internet at http://www.eppendorf.com
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service