Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 ea | Check Cart for Availability | € 597,00 |
About This Item
€ 597,00
material
polyethersulfone membrane
description
min recirculation volume < 20 mL
sterility
non-sterile
feature
holdup volume 5.3 mL
packaging
pkg of 1 ea
manufacturer/tradename
Sartorius VF20P0
availability
not available in ROW, USA and Canada
membrane area
200 cm2
working volume
200 μL , 500—5,000 mL
pore size
10 kDa MWCO
General description
1 of 1
This Item | |||
|---|---|---|---|
| description MWCO 10 kDa | description MWCO 100 kDa | description MWCO 5 kDa | description MWCO 50 kDa |
| manufacturer/tradename Sartorius VF20P0 | manufacturer/tradename Sartorius VF20P4 | manufacturer/tradename Sartorius VF20P1 | manufacturer/tradename Sartorius VF20P3 |
| pore size 10 kDa MWCO | pore size 100 kDa MWCO | pore size 5 kDa MWCO | pore size 50 kDa MWCO |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
| material polyethersulfone membrane | material polyethersulfone membrane | material polyethersulfone membrane | material polyethersulfone membrane |
| working volume 200 μL , 500—5,000 mL | working volume 200 μL , 500—5000 mL | working volume 200 μL , 500—5000 mL | working volume 200 μL , 500—5,000 mL |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
Still not finding the right product?
Explore all of our products under VivaFlow 200 flipflow filtration
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
