Z614033
Vivaspin 500 centrifugal concentrators
MWCO 10 kDa
All Photos(1)
About This Item
Recommended Products
material
polycarbonate housing
polyethersulfone membrane
description
PES membrane
sterility
non-sterile
feature
holdup volume 5 μL
packaging
package of 100 ea
manufacturer/tradename
Sartorius VS0102
parameter
15000 × g max. centrifugal force
dead stop volume
5 μL
membrane area
0.5 cm2
tube size
~2.2 mL
working volume
500 μL
pore size
10 kDa MWCO
General description
- Vivaspin centrifugal concentrators offer the optimal solution to any concentration or buffer exchange application with their broad range of volume capacities and MWCOs.
- Highest flow rates and recoveries are achieved due to their particular vertical membrane design.
- Incomparably safe sample handling and convenience is assured by the patented dead-stop technology allowing direct recovery of the concentrate without further re-spin.
Note: Vivaspin 500 μL centrifugal filter units offer a simple one step procedure for sample preparation. They can effectively be used in either swing out or fixed angle rotors.