Key Documents
Z601535
Hettich® centrifuge buckets
for ROTOFIX 32A, ROTINA 38/38R and UNIVERSAL 320/320R Centrifuges, bucket/carrier for rotors 1624; 1726
Select a Size
4 EA
€341.00
€341.00
Check Cart for Availability
Select a Size
Change View
4 EA
€341.00
About This Item
UNSPSC Code:
41103910
NACRES:
NB.22
€341.00
Check Cart for Availability
feature
Holds 7 x 15 mLtubes
packaging
pack of 4 ea
manufacturer/tradename
Hettich 1742
availability
available only in EU
not available in USA and Canada
technique(s)
centrifugation: suitable
compatibility
for use with rotors 1624 (Z601527) and 1726 (Z631108)
General description
  • compatible with rotors 1624 (Z601527) and 1726 (Z631108)
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
required but not provided
Choose from one of the most recent versions:
Certificates of Analysis (COA)