Key Documents
Z377910
Sigma-Aldrich
Agilent Technologies/Stratagene PCR microtubes
pkg of
Select a Size
1 PKG
€110.00
€110.00
Estimated to ship on13 May 2025
Select a Size
Change View
1 PKG
€110.00
About This Item
UNSPSC Code:
41121702
NACRES:
NB.22
€110.00
Estimated to ship on13 May 2025
material
polypropylene
description
Autoclavable
packaging
pkg of 120 strips (960 tubes)
pkg of
technique(s)
PCR: suitable
General description
Strips of eight caps can be cut apart to use individually.
Thin-walled, specifically designed to fit the RoboCycler96. Tested to ensure ideal contact with the thermal block; no mineral oil needed. Certified DNase- and RNase-free. All tolerate organic solvent reactions and temperatures from −4 to 121 °C.
Choose from one of the most recent versions:
Certificates of Analysis (COA)