Skip to Content
Merck
All Photos(1)

Key Documents

Z332593

Drum adapter system for BRAND® Dispensette® III and Dispensette® Organic

filling tube L × diam. 2.5 m × 830 mm, pore size 3 μm

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€265.00

€265.00


Check Cart for Availability


Select a Size

Change View
1 EA
€265.00

About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

€265.00


Check Cart for Availability

material

FEP filling tube

manufacturer/tradename

BRAND

filling tube L × diam.

2.5 m × 830 mm

pore size

3 μm

Related Categories

General description

  • Dispense accurate volumes directly from drums and bulk refills
  • The Dispensette can be mounted on a wall, a ring stand or on lab furniture
  • A filter in the drum adapter minimizes risk of contaminating high-purity reagents
  • A quick-release connector with integral valves allows quick changing of the bulk container
  • The remote dispensing system allows storage of the drum up to 10 m away from the Dispensette (the max delivery height is 1.2 m)
  • Standard supply (without Dispensette) for drum with 3/4 in. inner thread

Other Notes

BR704261 (India)

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service