Key Documents
Z332267
Bottle adapters for BRAND® Dispensers and Burettes
GL 32 thread, bottle (NS 24/29), polypropylene
Select a Size
1 EA
€11.90
€11.90
Estimated to ship on12 April 2025
Select a Size
Change View
1 EA
€11.90
About This Item
UNSPSC Code:
41121815
NACRES:
NB.01
€11.90
Estimated to ship on12 April 2025
material
polypropylene
manufacturer/tradename
BRAND 704424
fitting
bottle (NS 24/29)
GL 32 thread
General description
For all Dispensette® models as well as seripettor® and QuikSip
Other Notes
BR704424 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
QuikSip is a trademark of BRAND GMBH + CO KG
seripettor is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)