Key Documents
Z329738
BRAND® wash bottle, wide-mouth, labelled, LDPE
capacity 250 mL, label, Distilled water, without venting valve
Select a Size
5 EA
€33.50
€33.50
Check Cart for Availability
Select a Size
Change View
5 EA
€33.50
About This Item
UNSPSC Code:
41103209
NACRES:
NB.17
€33.50
Check Cart for Availability
material
low-density polyethylene bottle
natural bottle
round bottle
white closure
wide-mouth bottle
feature
label Distilled water
with cap
without venting valve
packaging
pack of 5 ea
manufacturer/tradename
BRAND 144078
capacity
250 mL
General description
Flexible, high-quality wash bottles with integral wash steam and sealing cone in the closure. Large opening for convenient filling. Label with designation of contents and general safety statements (with CAS no. and NFPA codes).
Bottles with venting valve (metal ball) are for distilled water and organic solvents only; to avoid dropping caused by overpressure.
Other Notes
BR144078 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
accessory
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)