Skip to Content
Merck
All Photos(1)

Key Documents

Z290807

Sigma-Aldrich

Nylon membrane filter discs.22 μm pore size

pore size 0.22 μm, diam. 47 mm, pack of 100

Synonym(s):

Nylon filter media, Nylon membrane filters

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
€184.00

€184.00


Estimated to ship on01 June 2025



Select a Size

Change View
100 EA
€184.00

About This Item

UNSPSC Code:
40161507
NACRES:
NB.24

€184.00


Estimated to ship on01 June 2025


Product Name

Nylon filter membranes, pore size 0.22 μm, diam. 47 mm, pack of 100

material

Nylon membrane

sterility

non-sterile

feature

hydrophilic

packaging

pack of 100

diam.

47 mm

thickness

60-125 μm

pore size

0.22 μm

Looking for similar products? Visit Product Comparison Guide

General description

Nylon filters are naturally hydrophilic and no wetting agents are used in manufacture.

With an extractable level <0.0015 mg/cm2, they are ideal for HPLC solvent and sample preparation. Because of high non-specific binding, they are not recommended for protein solutions.

Application

Nylon filter membranes have been used:

  • duringthe preparation of phosphate buffer saline (PBS)[1]
  • in the filtration ofraspberry fruit powder extract for liquid chromatography (LC)/tandem massspectrometry (MS/MS) analysis[2]
  • as a polymeric membrane to study itsinteraction with nanoparticles[3]

Features and Benefits

  • Compatible with aqueous and most organic solvents
  • Autoclave friendly
  • It can withstand temperatures up to 180°C

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alterations of phenolic compounds in red raspberry juice induced by high-hydrostatic-pressure and high-temperature short-time processing
Wentao Z, et al.
Innovative Food Science & Emerging Technologies, 67, 102569-102569 (2021)
Functionalized nanoparticle interactions with polymeric membranes
Ladner D A, et al.
Journal of Hazardous Materials, 211-212, 88-295 (2012)
Francesco Canfarotta et al.
Nature protocols, 11(3), 443-455 (2016-02-13)
Molecularly imprinted polymers (MIPs) are synthetic materials, generally based on acrylic or methacrylic monomers, that are polymerized in the presence of a specific target molecule called the 'template' and capable of rebinding selectively to this target molecule. They have the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service