Skip to Content
Merck
All Photos(1)

Key Documents

Y1751

Sigma-Aldrich

Yeast Synthetic Drop-out Medium Supplements

without histidine

Sign Into View Organizational & Contract Pricing

Select a Size

20 G
€186.00

€186.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
20 G
€186.00

About This Item

MDL number:
UNSPSC Code:
12000000
NACRES:
ND.02

€186.00


Estimated to ship on23 May 2025


Request a Bulk Order

Quality Level

form

powder

application(s)

food and beverages

Looking for similar products? Visit Product Comparison Guide

Application

The selection of plasmids in yeast is based on the use of auxotrophic mutant strains that cannot grow without a specific media component (an amino acid, purine or pyrimidine). Transformation with a plasmid containing the mutated gene enables the transformant to grow on a medium lacking the required component. Sigma′s Yeast Synthetic Drop-Out Media Supplements create a richer medium for better yield and growth rate, and increase the probability of successful transformations when screening libraries or performing gene knock-outs.

Components

The supplements contain all of the following components except as indicated:
Amino acids: All standard amino acids except for histidine. All the amino acids are present at a concentration of 76 mg/lL except for leucine, which is present at 380 mg/L.
Other nutrients: Adenine (18 mg/L), inositol (76 mg/L), uracil (76 mg/L), p-aminobenzoic acid (8 mg/L).

Other Notes

Mixtures of amino acids and other nutrients to be added to Yeast Nitrogen Base Without Amino Acids.

Quantity

1.92 g will supplement 1 liter of medium.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tamara Suhm et al.
Cell metabolism, 27(6), 1309-1322 (2018-05-15)
Cellular proteostasis is maintained via the coordinated synthesis, maintenance, and breakdown of proteins in the cytosol and organelles. While biogenesis of the mitochondrial membrane complexes that execute oxidative phosphorylation depends on cytoplasmic translation, it is unknown how translation within mitochondria impacts

Articles

Genetic engineering enables large-scale expression and isolation of recombinant proteins for research purposes.

Protocols

Magnetic-plasmonic nanoparticles: Synthesis and application of Ag/FeCo/Ag nanobeads for cellular organelle imaging and isolation.

Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service