Skip to Content
Merck
All Photos(5)

Key Documents

X3627

Sigma-Aldrich

Xanthine sodium salt

suitable for cell culture, ≥99%

Synonym(s):

2,6-Dihydroxypurine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€96.80
5 G
€365.00
10 G
€575.00
25 G
€1,290.00

€96.80


Estimated to ship on27 March 2025


Request a Bulk Order

Select a Size

Change View
1 G
€96.80
5 G
€365.00
10 G
€575.00
25 G
€1,290.00

About This Item

Empirical Formula (Hill Notation):
C5H3N4NaO2
CAS Number:
Molecular Weight:
174.09
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

€96.80


Estimated to ship on27 March 2025


Request a Bulk Order

biological source

synthetic (organic)

Quality Level

Assay

≥99%

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

≤4% methanol

solubility

1 M NaOH: 50 mg/mL, clear to slightly hazy, colorless to light yellow

SMILES string

[Na].O=C1NC(=O)c2[nH]cnc2N1

InChI

1S/C5H4N4O2.Na.H/c10-4-2-3(7-1-6-2)8-5(11)9-4;;/h1H,(H3,6,7,8,9,10,11);;

InChI key

JAZQMXVZANZJSO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Xanthine is commonly present in tea, coffee and cocoa.[1] Xanthine is a nitrogenous compound present in blood and urine.[2]

Application

Xanthine sodium salt has been used:
  • as a component in superoxide dismutase (SOD)-inhibitable ferricytochrome c reduction assay[3]
  • as a component in selection of chimeric Modified Vaccinia virus Ankara (MVA)[4]
  • as a substrate in the reaction mixture for xanthine oxidase assay[5]

Biochem/physiol Actions

Xanthine possesses therapeutic properties. It acts as a mild stimulant of the central nervous system.[1]

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 2

Target Organs

Eyes

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modern Pharmacology with Clinical Applications, 351-351 (2004)
Rau's Respiratory Care Pharmacology - E-Book (2013)
Uric acid-lowering effect and intestinal permeability of Kampo medicine, Hachimijiogan, Yokuininto and Goshakusan
Lee SH, et al.
European journal of integrative medicine, 20(1), 193-199 (2018)
Construction of FWPV Chimaeric MVA
Buttigieg KR and Skinner MA
Bio-protocol, 5(1), 65-80 (2015)
Assays for detecting biological superoxide
Zhu H
Reactive Oxygen Species, 1(1), 65-80 (2016)

Articles

Uric acid present in animal sera impacts serum-supplemented classical cell culture systems, influencing cell culture performance.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service