Skip to Content
Merck

WS80

Wright Stain, Modified

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116121
NACRES:
NA.47
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Quality Segment

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

0.3%

pH

6.75-6.95 (25 °C)

application(s)

hematology
histology

storage temp.

room temp

Application

Popular hematology stain used for differentially staining the cellular elements of blood. For dip, rack, and batch staining techniques.

Other Notes

Wright stain, 0.3%, buffered at pH 6.8 in methanol. Contains stabilizers and surfactant.


Still not finding the right product?

Explore all of our products under Wright Stain, Modified


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

91.4 °F - closed cup

flash_point_c

33 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

67-56-1

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Learn about the clinical study of blood, blood-forming organs, and blood diseases including the treatment, prevention, and stains and dyes used in hematology testing.

Informationen über die klinische Untersuchung von Blut, blutbildenden Organen und Blutkrankheiten, einschließlich deren Behandlung und Prävention, sowie über Farbstoffe und Färbemittel für die Hämatologie.





Global Trade Item Number

SKUGTIN
WS80-2.5L04061837543685