Skip to Content
Merck
All Photos(4)

Key Documents

WH0256297M5

Sigma-Aldrich

Monoclonal Anti-PTF1A antibody produced in mouse

clone 1A2, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-PTF1p48, Anti-pancreas specific transcription factor, 1a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1A2, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2a

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTF1A(256297)

General description

This gene encodes a protein that is a component of the pancreas transcription factor 1 complex (PTF1) and is known to have a role in mammalian pancreatic development. The protein plays a role in determining whether cells allocated to the pancreatic buds continue towards pancreatic organogenesis or revert back to duodenal fates. The protein is thought to be involved in the maintenance of exocrine pancreas-specific gene expression including elastase 1 and amylase. Mutations in this gene cause cerebellar agenesis and loss of expression is seen in ductal type pancreas cancers. (provided by RefSeq)

Immunogen

PTF1A (NP_835455, 250 a.a. ~ 328 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
QAQKVIICHRGTRSPSPSDPDYGLPPLAGHSLSWTDEKQLKEQNIIRTAKVWTPEDPRKLNSKSSFNNIENEPPFEFVS

Biochem/physiol Actions

Pancreas specific transcription factor, 1A (PTF1A) aids in the growth and differentiation of pancreas. It also has a control over excitatory and inhibitory neurons in the brain. Mutations in the PTF1A gene have been linked to cerebellar agenesis.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiple transcriptional mechanisms control Ptf1a levels during neural development including autoregulation by the PTF1-J complex.
Meredith DM
The Journal of Neuroscience, 29(36), 11139-11148 (2009)
ICAT is a novel Ptf1a interactor that regulates pancreatic acinar differentiation and displays altered expression in tumours.
Campos ML
BioChemistry: An Indian Journal, 451(30, 395-405 (2013)
A Turkish newborn infant with cerebellar agenesis/neonatal diabetes mellitus and PTF1A mutation.
Tutak E
Genetic Counseling (Geneva, Switzerland), 20(2), 147-152 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service