Skip to Content
Merck
All Photos(3)

Key Documents

WH0221120M1

Sigma-Aldrich

Monoclonal Anti-DEPC-1 antibody produced in mouse

clone 2A5-4F5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-prostate cancer antigen-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€492.00

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€492.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2A5-4F5, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ALKBH3(221120)

General description

The Escherichia coli AlkB protein protects against the cytotoxicity of methylating agents by repair of the specific DNA lesions generated in single-stranded DNA. ALKBH2 (MIM 610602) and ALKBH3 are E. coli AlkB homologs that catalyze the removal of 1-methyladenine and 3-methylcytosine (Duncan et al., 2002 [PubMed 12486230]).[supplied by OMIM

Immunogen

DEPC-1 (AAH15155, 1 a.a. ~ 139 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MEPNPHWHPVLRTLKNRIEENTGHTFNSLLCNLYRNEKDSVDWHSDDEPSLGRCPIIASLSFGATRTFEMRKKPPPEENGDYTYVERVKIPLDHGTLLIMEGATQADWQHRVPKEYHSREPRVNLTFRTVYPDPRGAPW

Biochem/physiol Actions

DEPC-1 is a DNA/RNA demethylase which has a high affinity for single-stranded DNA and RNA. It has been shown to demethylate N1-methyladenosine in mRNA and tRNA in vitro. DEPC-1 has a role in the development of prostate cancer and has been linked to urothelial carcinomas.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoyu Li et al.
Nature chemical biology, 12(5), 311-316 (2016-02-11)
N(1)-Methyladenosine (m(1)A) is a prevalent post-transcriptional RNA modification, yet little is known about its abundance, topology and dynamics in mRNA. Here, we show that m(1)A is prevalent in Homo sapiens mRNA, which shows an m(1)A/A ratio of ∼0.02%. We develop
Dan Dominissini et al.
Nature, 530(7591), 441-446 (2016-02-11)
Gene expression can be regulated post-transcriptionally through dynamic and reversible RNA modifications. A recent noteworthy example is N(6)-methyladenosine (m(6)A), which affects messenger RNA (mRNA) localization, stability, translation and splicing. Here we report on a new mRNA modification, N(1)-methyladenosine (m(1)A), that
Keiji Shimada et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 18(19), 5247-5255 (2012-08-02)
The role and function of a novel human AlkB homologue, ALKBH3, in human urothelial carcinoma development were examined. Biologic roles of ALKBH3 were examined by gene silencing analysis using in vitro and in vivo siRNA transfection. Immunohistochemical analyses of ALKBH3
Kiyohiko Hotta et al.
Oncology reports, 34(2), 648-654 (2015-06-03)
Prostate cancer antigen-1 (PCA-1)/ALKBH3 has been recently identified in human prostate cancer and its expression is correlated with disease progression and prognosis. However, the precise role and function of PCA-1/ALKBH3 in human malignancies are largely unknown. In the present study

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service