Skip to Content
Merck
All Photos(1)

Key Documents

WH0079092M1

Sigma-Aldrich

Monoclonal Anti-CARD14 antibody produced in mouse

clone 4B3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-BIMP2, Anti-CARMA2, Anti-caspase recruitment domain family, member 14

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€492.00

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€492.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4B3, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable

isotype

IgG2bκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CARD14(79092)

General description

The protein encoded by this gene belongs to the membrane-associated guanylate kinase (MAGUK) family, a class of proteins that functions as molecular scaffolds for the assembly of multiprotein complexes at specialized regions of the plasma membrane. This protein is also a member of the CARD protein family, which is defined by carrying a characteristic caspase-associated recruitment domain (CARD). This protein shares a similar domain structure with CARD11 protein. The CARD domains of both proteins have been shown to specifically interact with BCL10, a protein known to function as a positive regulator of cell apoptosis and NF-kappaB activation. When expressed in cells, this protein activated NF-kappaB and induced the phosphorylation of BCL10. Two alternatively spliced variants of this gene encoding distinct isoforms have been reported. (provided by RefSeq)

Immunogen

CARD14 (NP_077015, 905 a.a. ~ 1004 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
VQLDSVCTLHRMDIFPIVIHVSVNEKMAKKLKKGLQRLGTSEEQLLEAARQEEGDLDRAPCLYSSLAPDGWSDLDGLLSCVRQAIADEQKKVVWTEQSPR

Biochem/physiol Actions

The gene CARD14 (caspase recruitment domain family member 14) encodes a nuclear factor that contains a CARD domain involved in binding to the CARD domain of BCL10 (B cell leukemia 10), a protein involved in the activation of NF-κB through the IKK complex, and positive regulation of apoptosis. Mutations in this gene are associated with familial pityriasis rubra pilaris, a papulosquamous disorder phenotypically related to psoriasis. It also activates p38 and JNK MAP (c-Jun N-terminal kinase mitogen-activated protein) kinase pathways via association with paracaspase-1 (PCASP-1), also referred to as MALT1, which can serve as a therapeutic target in psoriasis.[1]

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Familial pityriasis rubra pilaris is caused by mutations in CARD14.
Fuchs-Telem D
American Journal of Human Genetics, 91, 163-170 (2012)
J Bertin et al.
The Journal of biological chemistry, 276(15), 11877-11882 (2001-03-30)
The caspase recruitment domain (CARD) is a protein-binding module that mediates the assembly of CARD-containing proteins into apoptosis and NF-kappaB signaling complexes. We report here that CARD protein 11 (CARD11) and CARD protein 14 (CARD14) are novel CARD-containing proteins that
Inna S Afonina et al.
EMBO reports, 17(6), 914-927 (2016-04-27)
Mutations in CARD14 have recently been linked to psoriasis susceptibility. CARD14 is an epidermal regulator of NF-κB activation. However, the ability of CARD14 to activate other signaling pathways as well as the biochemical mechanisms that mediate and regulate its function

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service