Skip to Content
Merck
All Photos(4)

Key Documents

WH0079058M1

Sigma-Aldrich

Monoclonal Anti-ASPSCR1 antibody produced in mouse

clone 3D10-1D11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-ASPCR1, Anti-ASPL, Anti-ASPS, Anti-RCC17, Anti-TUG, Anti-alveolar soft part sarcoma chromosome region, candidate 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€492.00

€492.00


Estimated to ship on27 April 2025



Select a Size

Change View
100 μG
€492.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€492.00


Estimated to ship on27 April 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3D10-1D11, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ASPSCR1(79058)

General description

This gene is a candidate gene for alveolar soft part sarcoma (ASPS). It has been found that this gene is fused with transcription factor TFE3 gene in ASPS and also in renal cell carcinomas. Several alternatively spliced transcript variants of this gene have been described, but their full length nature has not been determined. (provided by RefSeq)

Immunogen

ASPSCR1 (AAH18722, 1 a.a. ~ 553 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MAAPAGGGGSAVSVLAPNGRRHTVKVTPSTVLLQVLEDTCRRQDFNPCEYDLKFQRSVLDLSLQWRFANLPNNAKLEMVPASRSREGPENMVRIALQLDDGSRLQDSFCSGQTLWELLSHFPQIRECLQHPGGATPVCVYTRDEVTGEAALRGTTLQSLGLTGGSATIRFVMKCYDPVGKTPGSLGSSASAGQAAASAPLPLESGELSRGDLSRPEDADTSGPCCEHTQEKQSTRAPAAAPFVPFSGGGQRQGGPPGPTRPLTSSSAKLPKSLSSPGGPSKPKKSKSGQDPQQEQEQERERDPQQEQERERPVDREPVDREPVVCHPDLEERLQAWPAELPDEFFELTVDDVRRRLAQLKSERKRLEEAPLVTKAFREAQIKEKLERYPKVALRVLFPDRYVLQGFFRPSETVGDLRDFVRSHLGNPELSFYLFITPPKTVLDDHTQTLFQANLFPAALVHLGAEEPAGVYLEPGLLEHAISPSAADVLVARYMSRAAGSPSPLPAPDPAPKSEPAAEEGALVPPEPIPGTAQPVKRSLGKVPKWLKLPASKR

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peng Huang et al.
FEBS open bio, 13(11), 2094-2107 (2023-09-21)
Glucose transporters (GLUTs) are responsible for transporting hexose molecules across cellular membranes. In adipocytes, insulin stimulates glucose uptake by redistributing GLUT4 to the plasma membrane. In unstimulated adipose-like mouse cell lines, GLUT4 is known to be retained intracellularly by binding
Alexandra Kuzyk et al.
Genes, chromosomes & cancer, 54(10), 616-628 (2015-07-15)
MYCN amplification and MYCN overexpression are poor prognostic factors in neuroblastoma. Tumors with unbalanced chromosome arm 17q gain are often associated with MYCN amplification; however, the relationship between chromosome 17 copy number status and MYCN expression is not known. We

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service