Skip to Content
Merck
All Photos(3)

Key Documents

WH0055507M1

Sigma-Aldrich

Monoclonal Anti-GPRC5D antibody produced in mouse

clone 6D9, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-G protein-coupled receptor, family C, group 5, member D

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€492.00

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€492.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6D9, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GPRC5D(55507)

General description

G protein-coupled receptor 5D (GPRC5D), a surface receptor consists of seven putative transmembrane segments. It is expressed in the cell membrane. This protein belongs to the retinoic acid inducible gene 1 or retinoic acid inducible GPCR 1 (RAIG1) family. GPRC5D is located on human chromosome 12p13.

Immunogen

GPRC5D (NP_061124, 261 a.a. ~ 345 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
ELCILYRSCRQECPLQGNACPVTAYQHSFQVENQELSRARDSDGAEEDVALTSYGTPIQPQTVDPTQECFIPQAKLSPQQDAGGV

Biochem/physiol Actions

G protein-coupled receptor 5D (GPRC5D) considered as an important marker for monitoring the tumor load and also to target multiple myeloma cells. Overexpression of GPRC5D affects the expression of hard keratins and also decrease the metabolic activities of hair bulb cells (HBCs).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

GPRC5D is a promising marker for monitoring the tumor load and to target multiple myeloma cells
Cohen Y, et al.
Hematology, 18(6), 348-351 (2013)
Cloning and characterization of a human orphan family C G-protein coupled receptor GPRC5D
Brauner-OH, et al.
Biochimica et Biophysica Acta (BBA)-Gene Structure and Expression, 1518(3), 237-248 (2001)
The RAIG family member, GPRC5D, is associated with hard-keratinized structures
Inoue S, et al.
The Journal of Investigative Dermatology, 122(3), 565-573 (2004)
Overexpression of G protein-coupled receptor 5D in the bone marrow is associated with poor prognosis in patients with multiple myeloma
Atamaniuk J, et al.
European Journal of Clinical Investigation, 42(9), 953-960 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service