WH0022932M2
Monoclonal Anti-POMZP3 antibody produced in mouse
clone 2E7, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Anti-MGC8359, Anti-POM (POM121 homolog, rat) and ZP3 fusion, Anti-POM121, Anti-POMZP3
Select a Size
€492.00
Select a Size
About This Item
€492.00
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
2E7, monoclonal
form
buffered aqueous solution
species reactivity
rat, mouse, human
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL
isotype
IgG1κ
GenBank accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... POMZP3(22932)
Related Categories
General description
Immunogen
Sequence
DQIFLDGQENKRSCLVDGLTDASSAFKVPRPGPDTLQFTVDVFHFANDSRNM*
Physical form
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service