WH0010629M1
Monoclonal Anti-TAF6L antibody produced in mouse
clone 1C5, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Anti-FLJ11136, Anti-MGC4288, Anti-PAF65A, Anti-TAF6-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associated factor, 65kDa
About This Item
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
1C5, monoclonal
form
buffered aqueous solution
species reactivity
human
technique(s)
immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL
isotype
IgG2bκ
GenBank accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... TAF6L(10629)
General description
Immunogen
Sequence
SEREERRFVEIPRESVRLMAESTGLELSDEVAALLAEDVCYRLREATQNSSQFMKHTKRRKLTVEDFNRALRWSSVEAVCGYGSQEALPMRPAREGELYFPEDREVNL
Physical form
Legal Information
Not finding the right product?
Try our Product Selector Tool.
recommended
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service