Skip to Content
Merck
All Photos(4)

Key Documents

WH0010611M1

Sigma-Aldrich

Monoclonal Anti-PDLIM5 antibody produced in mouse

clone 3E11-F6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-ENH, Anti-L9, Anti-LIM, Anti-PDZ and LIM domain 5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€418.00

€418.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€418.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€418.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3E11-F6, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDLIM5(10611)

General description

The protein encoded by this gene is a LIM domain protein. LIM domains are cysteine-rich double zinc fingers composed of 50 to 60 amino acids that are involved in protein-protein interactions. LIM domain-containing proteins are scaffolds for the formation of multiprotein complexes. The proteins are involved in cytoskeleton organization, cell lineage specification, organ development, and oncogenesis. The encoded protein is also a member of the Enigma class of proteins, a family of proteins that possess a 100-amino acid PDZ domain in the N terminus and 1 to 3 LIM domains in the C terminus. Multiple transcript variants encoding different isoforms have been found for this gene, although not all of them have been fully characterized. (provided by RefSeq)

Immunogen

PDLIM5 (AAH08741, 1 a.a. ~ 596 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MSNYSVSLVGPAPWGFRLQGGKDFNMPLTISSLKDGGKAAQANVRIGDVVLSIDGINAQGMTHLEAQNKIKGCTGSLNMTLQRASAAPKPEPVPVQKGEPKEVVKPVPITSPAVSKVTSTNNMAYNKAPRPFGSVSSPKVTSIPSPSSAFTPAHATTSSHASPSPVAAVTPPLFAASGLHANANLSADQSPSALSAGKTAVNVPRQPTVTSVCSETSQELAEGQRRGSQGDSKQQNGPPRKHIVERYTEFYHVPTHSDASKKRLIEDTEDWRPRTGTTQSRSFRILAQITGTEHLKESEADNTKKANNSQEPSPQLASSVASTRSMPESLDSPTSGRPGVTSLTTAAAFKPVGSTGVIKSPSWQRPNQGVPSTGRISNSAAYSGSVAPANSALGQTQPSDQDTLVQRAEHIPAGKRTPMCAHCNQVIRGPFLVALGKSWHPEEFNCAHCKNTMAYIGFVEEKGALYCELCYEKFFAPECGRCQRKILGEVINALKQTWHVSCFVCVACGKPIRNNVFHLEDGEPYCETDYYALFGTICHGCEFPIEAGDMFLEALGYTWHDTCFVCSVCCESLEGQTFFSKKDKPLCKKHAHSVNF

Biochem/physiol Actions

PDLIM5 (PDZ and LIM domain 5) is involved in the several cellular processes such as cytoskeleton organization, cell lineage specification, organ development, and oncogenesis. It also acts as a scaffold protein that helps to link protein kinases and transcription factors with cytoskeletal elements. Nonsynonymous polymorphisms of the PDLIM5 gene cause both bipolar disorder (BPD) and schizophrenia.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yogesh Srivastava et al.
Frontiers in cellular neuroscience, 18, 1315941-1315941 (2024-02-28)
Neuronal connectivity is regulated during normal brain development with the arrangement of spines and synapses being dependent on the morphology of dendrites. Further, in multiple neurodevelopmental and aging disorders, disruptions of dendrite formation or shaping is associated with atypical neuronal
Ryan Baumert et al.
The Journal of cell biology, 219(11) (2020-10-03)
In neurons, dendrites form the major sites of information receipt and integration. It is thus vital that, during development, the dendritic arbor is adequately formed to enable proper neural circuit formation and function. While several known processes shape the arbor
Mohd A Zain et al.
Psychiatric genetics, 23(6), 258-261 (2013-09-26)
Two single nucleotide polymorphisms of PDLIM5, rs7690296 and rs11097431, were genotyped using Mass-Array SNP genotyping by Sequenom technology in 244 bipolar disorder patients, 471 schizophrenia patients, and 601 control individuals who were Malay, Chinese, and Indian ethnic groups in the
Andrés D Maturana et al.
Cellular signalling, 23(5), 928-934 (2011-01-27)
Enigma homolog protein 1 (ENH1) acts as a scaffold that selectively associates protein kinases and transcription factors with cytoskeletal elements. ENH1 comprises an N-terminal PDZ domain and three C-terminal LIM domains. Through the LIM domains ENH1 interacts with the N-terminal
Peiheng Gan et al.
Developmental cell, 57(8), 959-973 (2022-04-27)
Noncompaction cardiomyopathy is a common congenital cardiac disorder associated with abnormal ventricular cardiomyocyte trabeculation and impaired pump function. The genetic basis and underlying mechanisms of this disorder remain elusive. We show that the genetic deletion of RNA-binding protein with multiple

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service