Skip to Content
Merck
All Photos(5)

Key Documents

WH0004999M1

Sigma-Aldrich

Monoclonal Anti-ORC2L antibody produced in mouse

clone 3E11-1G5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-ORC2, Anti-origin recognition complex, subunit 2-like (yeast)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€492.00

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€492.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3E11-1G5, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ORC2L(4999)

General description

The origin recognition complex (ORC) is a highly conserved six subunits protein complex essential for the initiation of the DNA replication in eukaryotic cells. Studies in yeast demonstrated that ORC binds specifically to origins of replication and serves as a platform for the assembly of additional initiation factors such as Cdc6 and Mcm proteins. The protein encoded by this gene is a subunit of the ORC complex. This protein forms a core complex with ORC3L, -4L, and -5L. It also interacts with CDC45L and MCM10, which are proteins known to be important for the initiation of DNA replication. This protein has been demonstrated to specifically associate with the origin of replication of Epstein-Barr virus in human cells, and is thought to be required for DNA replication from viral origin of replication. (provided by RefSeq)

Immunogen

ORC2L (AAH14834, 1 a.a. ~ 577 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MSKPELKEDKMLEVHFVGDDDVLNHILDREGGAKLKKERAQLLVNPKKIIKKPEYDLEEDDQEVLKDQNYVEIMGRDVQESLKNGSATGGGNKVYSFQNRKHSEKMAKLASELAKTPQKSVSFSLKNDPEITINVPQSSKGHSASDKVQPKNNDKSEFLSTAPRSLRKRLIVPRSHSDSESEYSASNSEDDEGVAQEHEEDTNAVIFSQKIQAQNRVVSAPVGKETPSKRMKRDKTSDLVEEYFEAHSSSKVLTSDRTLQKLKRAKLDQQTLRNLLSKVSPSFSAELKQLNQQYEKLFHKWMLQLHLGFNIVLYGLGSKRDLLERFRTTMLQDSIHVVINGFFPGISVKSVLNSITEEVLDHMGTFRSILDQLDWIVNKFKEDSSLELFLLIHNLDSQMLRGEKSQQIIGQLSSLHNIYLIASIDHLNAPLMWDHAKQSLFNWLWYETTTYSPYTEETSYENSLLVKQSGSLPLSSLTHVLRSLTPNARGIFRLLIKYQLDNQDNPSYIGLSFQDFYQQCREAFLVNSDLTLRAQLTEFRDHKLIRTKKGTDGVEYLLIPVDNGTLTDFLEKEEEEA

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service