Skip to Content
Merck
All Photos(2)

Key Documents

WH0002895M1

Sigma-Aldrich

Monoclonal Anti-GRID2 antibody produced in mouse

clone 1A1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-MGC117022, Anti-MGC117023, Anti-MGC117024, Anti-glutamate receptor, ionotropic, delta 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€492.00

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€492.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€492.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1A1, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GRID2(2895)

General description

Human glutamate receptor delta-2 (GRID2) is a relatively new member of the family of ionotropic glutamate receptors which are the predominant excitatory neurotransmitter receptors in the mammalian brain. GRID2 is a predicted 1,007 amino acid protein that shares 97% identity with the mouse homolog which is expressed selectively in cerebellar Purkinje cells. A point mutation in mouse GRID2, associated with the phenotype named ′lurcher′, in the heterozygous state leads to ataxia resulting from selective, cell-autonomous apoptosis of cerebellar Purkinje cells during postnatal development. Mice homozygous for this mutation die shortly after birth from massive loss of mid- and hindbrain neurons during late embryogenesis. This strongly suggests a role for GRID2 in neuronal apoptotic death. (provided by RefSeq)

Immunogen

GRID2 (NP_001501, 908 a.a. ~ 1007 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
DTLPTRQALEQISDFRNTHITTTTFIPEQIQTLSRTLSAKAASGFTFGNVPEHRTGPFRHRAPNGGFFRSPIKTMSSIPYQPTPTLGLNLGNDPDRGTSI

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service