Skip to Content
Merck
All Photos(2)

Key Documents

WH0002697M1

Sigma-Aldrich

Monoclonal Anti-GJA1 antibody produced in mouse

clone 3E5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CX43, Anti-DFNB38, Anti-GJAL, Anti-ODD, Anti-ODDD, Anti-ODOD, Anti-SDTY3, Anti-gap junction protein, alpha 1, 43kDa (connexin 43)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3E5, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... GJA1(2697)

General description

This gene is a member of the connexin gene family. The encoded protein is a component of gap junctions, which are composed of arrays of intercellular channels that provide a route for the diffusion of low molecular weight materials from cell to cell. The encoded protein is the major protein of gap junctions in the heart that are thought to have a crucial role in the synchronized contraction of the heart and in embryonic development. A related intronless pseudogene has been mapped to chromosome 5. Mutations in this gene have been associated with oculodentodigital dysplasia and heart malformations. (provided by RefSeq)

Immunogen

GJA1 (AAH26329, 261 a.a. ~ 361 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
GSQKYAYFNGCSSPTAPLSPMSPPGYKLVTGDRNNSSCRNYNKQASEQNWANYSAEQNRMGQAGSTISNSHAQPFDFPDDNQNSKKLAAGHELQPLAIVD*

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xi Xie et al.
Experimental cell research, 327(2), 276-286 (2014-07-30)
RhoA/Rho kinase (ROCK) signaling has been suggested to be involved in diabetic nephropathy (DN) pathogenesis. Altered expression of connexin43 (Cx43) has been found in kidneys of diabetic animals. Both of them have been found to regulate nuclear factor kappa-B (NF-κB)
Chang-Qing Fan et al.
International heart journal, 55(4), 362-371 (2014-06-27)
Impact of early bone marrow-derived mesenchymal stem cell (BMDMSC) implantation on left ventricular (LV) function after AMI was studied.Twelve mini-pigs were equally divided into placebo (AMI through left coronary artery ligation) and cell-treated groups [BMDMSCs (3.0 × 10(7)) implanted into
Endika Haro et al.
PLoS genetics, 10(8), e1004468-e1004468 (2014-08-29)
The formation and maintenance of the apical ectodermal ridge (AER) is critical for the outgrowth and patterning of the vertebrate limb. The induction of the AER is a complex process that relies on integrated interactions among the Fgf, Wnt, and
Yuanyuan Xue et al.
Journal of cellular biochemistry, 115(9), 1495-1504 (2014-03-13)
Insulin-like growth factor binding protein 4 (IGFBP4) has been reported to play critical role in cardiomyocytes differentiation of embryonic stem cells (ESCs). But whether it promotes cardiomyocytes induction of iPSCs is unclear. In the present study, we aim to explore
Cindy X Guo et al.
Molecular vision, 20, 670-682 (2014-06-03)
Changes in connexin expression are associated with many pathological conditions seen in animal models and in humans. We hypothesized that gap junctions are important mediators in tissue dysfunction and injury processes in the retina, and therefore, we investigated the pattern

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service