Skip to Content
Merck
All Photos(7)

Key Documents

WH0001027M1

Sigma-Aldrich

Monoclonal Anti-CDKN1B antibody produced in mouse

clone 4B4-E6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CDKN4, Anti-KIP1, Anti-P27KIP1, Anti-cyclin-dependent kinase inhibitor 1B (p27, Kip1)

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€411.00

€411.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB1429


Select a Size

Change View
50 μG
€411.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€411.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB1429

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4B4-E6, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

ELISA: suitable
capture ELISA: suitable
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
proximity ligation assay: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDKN1B(1027)

General description

This gene encodes a cyclin-dependent kinase inhibitor, which shares a limited similarity with CDK inhibitor CDKN1A/p21. The encoded protein binds to and prevents the activation of cyclin E-CDK2 or cyclin D-CDK4 complexes, and thus controls the cell cycle progression at G1. The degradation of this protein, which is triggered by its CDK dependent phosphorylation and subsequent ubiquitination by SCF complexes, is required for the cellular transition from quiescence to the proliferative state. (provided by RefSeq)

Immunogen

CDKN1B (AAH01971, 1 a.a. ~ 198 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MSNVRVSNGSPSLERMDARQAEHPKPSACRNLFGPVDHEELTRDLEKHCRDMEEASQRKWNFDFQNHKPLEGKYEWQEVEKGSLPEFYYRPPRPPKGACKVPAQESQDGSGSRPAAPLIGAPANSEDTHLVDPKTDPSDSQTGLAEQCAGIRKRPATGDSSTQNKRANRTEENVSDGSPNAGSVEQTPKKPGLRRRQT

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yue Liu et al.
International journal of ophthalmology, 7(4), 594-601 (2014-08-28)
To discuss the prognostic significant of autophagy related proteins (ARPs) in retinoblastoma (RB) and to find the molecular marker to distinguish retinocytoma (RC) and RB by investigating the different expression profiling of microtubule-associated protein light chain 3 (LC3B) and other

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service