Skip to Content
Merck
All Photos(3)

Key Documents

WH0000324M1

Sigma-Aldrich

Monoclonal Anti-APC antibody produced in mouse

clone 3E2, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-DP2, Anti-DP2.5, Anti-DP3, Anti-FAP, Anti-FPC, Anti-GS, Anti-adenomatosis polyposis coli

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€418.00

€418.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€418.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€418.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3E2, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
proximity ligation assay: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... APC(324)

General description

Adenomatous polyposis coli protein (APC) is encoded by the gene mapped to human chromosome 5q. The encoded protein is composed of 2843 amino acids. APC exists as many transcripts and they are expressed in tissue specific manner.
This gene encodes a tumor suppressor protein that acts as an antagonist of the Wnt signaling pathway. It is also involved in other processes including cell migration and adhesion, transcriptional activation, and apoptosis. Defects in this gene cause familial adenomatous polyposis (FAP), an autosomal dominant pre-malignant disease that usually progresses to malignancy. Disease-associated mutations tend to be clustered in a small region designated the mutation cluster region (MCR) and result in a truncated protein product. (provided by RefSeq)

Immunogen

APC (NP_000029, 2744 a.a. ~ 2843 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
NPVPVSETNESSIVERTPFSSSSSSKHSSPSGTVAARVTPFNYNPSPRKSSADSTSARPSQIPTPVNNNTKKRDSKTDSTESSGTQSPKRHSGSYLVTSV

Biochem/physiol Actions

Adenomatous polyposis coli protein (APC) is a tumor suppressor gene. It plays an important role in cell division, adhesion and migration. Mutations in this gene are associated with familial adenomatous polyposis (FAP). APC is involved with Wnt pathway and inhibits the molecules which induce β-catenin expression, an agent participating in cellular transformation. It also regulates metaphase-anaphase transition, mitotic exit and G1 phase, thereby maintaining genomic stability and chromosome segregation.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adenomatous polyposis coli (APC) regulates miR17-92 cluster through ?-catenin pathway in colorectal cancer.
Li Y
Oncogene, 35, 4558-4568 (2016)
APC Mutations as a Potential Biomarker for Sensitivity to Tankyrase Inhibitors in Colorectal Cancer.
Tanaka N
Molecular Cancer Therapeutics, 16, 752-762 (2017)
Epigenetic regulation of APC in the molecular pathogenesis of gallbladder cancer.
Tekcham DS
The Indian Journal of Medical Research, S82-S90 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service