Skip to Content
Merck
All Photos(3)

Key Documents

V9139

Sigma-Aldrich

Anti-Visfatin (C-terminal) antibody produced in rabbit

~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-NAmPRTase, Anti-NMPT, Anti-Nicotinamide-phosphoribosyltransferase, Anti-PBEF1, Anti-Pre-B-cell colony enhancing factor 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~50 kDa

species reactivity

rat, human, mouse (predicted)

concentration

~1.5 mg/mL

technique(s)

western blot: 1-2 μg/mL using A549 cell lysate
western blot: 2-4 μg/mL using rat lung extract (S1 fraction)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NAMPT(10135)
mouse ... Nampt(59027)
rat ... Nampt(297508)

General description

The gene for visfatin (also known as pre-B colony enhancing factor 1, PBEF1, nicotinamide phosphoribosyl transferase, NAMPT), is mapped to human chromosome 7q22.3. The encoded protein is an adipokine produced from adipocytes. It is a highly conserved protein and is also secreted by lymphocytes. Visfatin is expressed in large amounts in the bone marrow, liver tissue, and muscle and is also present in heart, placenta, lung, and kidney tissues.

Specificity

Anti-Visfatin (C- terminal) recognizes human and rat visfatin.

Application

Anti-Visfatin (C-terminal) antibody produced in rabbit has been used in:
  • western blotting(10)
  • immunohistochemistry
  • immunofluorescence

Biochem/physiol Actions

Visfatin is an enzyme that catalyzes the biosynthesis of nicotinamide adeninine dinucleotide (NAD) from nicotinamide. A variety of signaling pathways are regulated by visfatin including phosphatidylinositol 3-kinase (PI3K)/protein kinase B (AKT), signal transducer and activator of transcription 3 (STAT3) and extracellular signal-regulated protein kinase (ERK1/2). It is also reported to regulate the activity of sirtuin 1 (SIRT1) and protect the mammalian cells from death. Since visfatin catalyses a major rate-limiting step in the production of NAD, it is crucial in metabolism, carcinogenesis and immune responses. Visfatin also can act as cytokine and regulates the expression of vascular endothelial growth factor (VEGF) in the proliferation of endothelial cells and formation of capillary tubes. Deregulation of visfatin has been implicated in diabetes mellitus, obesity, rheumatoid arthritis and chronic hepatitis
Visfatin is involved in the regulation of energy homeostasis. Visfatin has been shown to activate its target cells by binding to the insulin receptor at a site distinct from insulin, and its expression level in plasma increases during the development of obesity. Visfatin can induce the cellular expression of inflammatory cytokines such as tumor necrosis factor (TNF)-α, interleukin (IL)-1β, IL-7,(11) and IL-6.

Target description

Visfatin (C-terminal) (also known as pre-B colony enhancing factor1, PBEF1, nicotinamide phosphoribosyltransferase, NAMPT), is a highly conserved protein involved in the regulation of cellular metabolism and energy homeostasis.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dietary phytoestrogen diosgenin interrupts metabolism, physiology, and reproduction of Swiss albino mice: Possible mode of action as an emerging environmental contaminant, endocrine disruptor and reproductive toxicant.
Khushboo, et al.
Food And Chemical Toxicology, 176, 113798-113798 (2023)
Thai Hien Tu et al.
Mediators of inflammation, 2017, 1958947-1958947 (2018-01-25)
Visfatin is an adipokine that is secreted from adipose tissue, and it is involved in a variety of physiological processes. In particular, visfatin has been implicated in metabolic diseases, such as obesity and type 2 diabetes, which are directly linked
The Nicotinamide Phosphoribosyltransferase: A Molecular Link between Metabolism, Inflammation, and Cancer
Galli M et al
Cancer Research, 70, doi: 10-doi: 10 (2010)
E Wanecq et al.
Journal of physiology and biochemistry, 65(4), 351-359 (2010-04-02)
Visfatin, a protein identified as a secretion product of visceral fat in humans and mice, is also expressed in different anatomical locations, and is known as pre-B cell-colony enhancing factor (PEBF1). It is also an enzyme displaying nicotinamide phosphoribosyltransferase activity
Tracy Luk et al.
Journal of leukocyte biology, 83(4), 804-816 (2008-02-07)
Pre-B cell colony-enhancing factor (PBEF), also known as visfatin, is a highly conserved, 52-kDa protein found in living species from bacteria to humans. Originally a curiosity identified serendipitously in microarray studies but having no obvious functional importance, PBEF has now

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service