Tityus serrulatus scorpion venom (Tsv) may be used as a source of individual Tsv components, as an immunogen and for studies of its physiological and biochemical affects.
Storage Class Code
6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
Toxicon : official journal of the International Society on Toxinology, 60(8), 1349-1354 (2012-09-26)
Several species of scorpions are known to cause accidents which can lead to death, most of them belonging to the genus Tityus. Tityus serrulatus is considered the most dangerous scorpion in South America. In Brazil, T. serrulatus is responsible for
Protein and peptide letters, 16(8), 920-932 (2009-08-20)
Tityus serrulatus is considered the most dangerous scorpion in South America and responsible for most of the fatal cases. This review will focus on Tityus serrulatus scorpion venom (Tsv), its long-chain Na(+)-channel toxins (NaTx), which include alpha- and beta-neurotoxins, short-chain
Toxicon : official journal of the International Society on Toxinology, 56(8), 1477-1486 (2010-09-15)
The aim of this work was to evaluate the hematological changes induced by Tityus serrulatus venom (TsV). Blood of Wistar rats was collected 0.5, 2, 6 and 24 h after i.p. injection of TsV (0.5 mg/kg) or saline (controls). Two additional groups
Toxicon : official journal of the International Society on Toxinology, 58(6-7), 480-485 (2011-09-07)
Despite several studies showed that the Tityus serrulatus scorpion venom (Tsv) induces an inflammatory response, just a few have investigated the effect of the venom on the immune response. Therefore, the aim of this study was to evaluate alterations of
Toxicon : official journal of the International Society on Toxinology, 61, 1-10 (2012-10-23)
Inflammatory mediators are thought to be involved in the systemic and local immune response induced by the Tityus serrulatus scorpion envenomation. New functional aspects of lipid mediators have recently been described. Here, we examine the unreported role of lipid mediators
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.