Skip to Content
Merck
All Photos(1)

Key Documents

V1258

Sigma-Aldrich

[Arg8]-Vasotocin-Gly-Lys-Arg

≥97% (HPLC)

Synonym(s):

Hydrin I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H93N21O16S2
CAS Number:
Molecular Weight:
1392.61
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Product Name

[Arg8]-Vasotocin-Gly-Lys-Arg, ≥97% (HPLC)

Assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CCC(C)C1NC(=O)C(Cc2ccc(O)cc2)NC(=O)C(N)CSSCC(NC(=O)C(CC(N)=O)NC(=O)C(CCC(N)=O)NC1=O)C(=O)N3CCCC3C(=O)NC(CCCNC(N)=N)C(=O)NCC(=O)NCC(=O)NC(CCCCN)C(=O)NC(CCCNC(N)=N)C(O)=O

InChI

1S/C57H93N21O16S2/c1-3-29(2)45-53(91)72-35(17-18-41(60)80)49(87)75-38(24-42(61)81)50(88)76-39(28-96-95-27-32(59)46(84)74-37(51(89)77-45)23-30-13-15-31(79)16-14-30)54(92)78-22-8-12-40(78)52(90)71-33(10-6-20-66-56(62)63)47(85)69-25-43(82)68-26-44(83)70-34(9-4-5-19-58)48(86)73-36(55(93)94)11-7-21-67-57(64)65/h13-16,29,32-40,45,79H,3-12,17-28,58-59H2,1-2H3,(H2,60,80)(H2,61,81)(H,68,82)(H,69,85)(H,70,83)(H,71,90)(H,72,91)(H,73,86)(H,74,84)(H,75,87)(H,76,88)(H,77,89)(H,93,94)(H4,62,63,66)(H4,64,65,67)

InChI key

BJRAYGWQMPNTHC-UHFFFAOYSA-N

Gene Information

human ... AVP(551)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Acher et al.
Biology of the cell, 89(5-6), 283-291 (1997-08-01)
Volemia and osmolality homeostasis is ensured in vertebrates through neuroendocrine reflexes, involving an afferent neural branch from baro- and osmo-receptors to hypothalamus and an efferent endocrine branch from secretory neurons to target hydroosmotic cells equipped with receptors and effectors. Whereas
P Eggena et al.
The American journal of physiology, 259(4 Pt 1), E524-E528 (1990-10-01)
Hydrin 1 is the biosynthetic precursor of vasotocin in Xenopus laevis. We have synthesized deamino and fluorescein analogues of hydrin 1 and characterized their physiological action in the urinary bladder of the toad, Bufo marinus. 1-Deamino-hydrin 1 (d-hydrin) was more
J T Winslow et al.
European journal of pharmacology, 233(1), 101-107 (1993-03-16)
The neurohypophyseal peptide hormone arginine-vasopressin functions as a neuropeptide in several brain areas in addition to its role as a posterior pituitary hormone. Several studies indicate that arginine-vasopressin and arginine-vasopressin receptors appear early in the infant rat brain. To determine
K W Chiu et al.
General and comparative endocrinology, 91(1), 1-7 (1993-07-01)
Cardiovascular and renal actions of hydrins and arginine vasotocin (AVT) in frogs (Rana tigrina and Rana catesbeiana) were examined in view of their relationships as precursor/prohormone-hormones in the anuran amphibian. In vitro hydrins and AVT respectively vasorelaxed and vasoconstricted KCl-preconstricted
Y Rouille et al.
Neuropeptides, 29(5), 301-307 (1995-11-01)
The biological properties of vasotocin, hydrin 1 (vasotocinyl-Gly-Lys-Arg) and hydrin 2 (vasotocinyl-Gly), in particular the hydro-osmotic activities on the frog skin, the frog urinary bladder and the frog kidney, have been compared. Hydrins are as active or more active than

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service