Skip to Content
Merck
All Photos(1)

Key Documents

U9257

Sigma-Aldrich

UbcH9 Conjugating Enzyme human

≥95% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€591.00

€591.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€591.00

About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.56

€591.00


Check Cart for Availability

Request a Bulk Order

recombinant

expressed in E. coli

Quality Level

Assay

≥95% (SDS-PAGE)

form

buffered aqueous glycerol solution

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Manufactured for Sigma by Boston Biochem., Inc.

Biochem/physiol Actions

UbcH9 mediates the conjugation of SUMO-1, SUMO-2, and SUMO-3 to a diverse variety of proteins without the help of an E3 ligase. This E2 conjugating enzyme interacts directly with protein substrates that are modified by SUMOylation, and may help in substrate recognition. UbcH9 is specific for the conjugation of proteins modified by SUMOylation.

Physical form

Solution in 50 mM HEPES, pH 7.5, 100 mM NaCl, 10 % glycerol, 1mM TCEP.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M Desterro et al.
FEBS letters, 417(3), 297-300 (1997-12-31)
Ubiquitin conjugating enzymes participate in the thioester cascade that leads to protein ubiquitination. Although Ubc9 is homologous to E2 ubiquitin conjugating enzymes we have shown that it is unable to form a thioester with ubiquitin, but can form a thioester

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service