Skip to Content
Merck
All Photos(3)

Key Documents

U6381

Sigma-Aldrich

Uridine

BioUltra, ≥99%

Synonym(s):

1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€160.00
25 G
€538.00

€160.00


Estimated to ship on13 April 2025



Select a Size

Change View
5 G
€160.00
25 G
€538.00

About This Item

Empirical Formula (Hill Notation):
C9H12N2O6
CAS Number:
Molecular Weight:
244.20
Beilstein:
754902
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.26

€160.00


Estimated to ship on13 April 2025


biological source

synthetic (organic)

Quality Level

product line

BioUltra

Assay

≥99%

form

powder

impurities

≤0.016% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.2%

mp

163-167 °C (lit.)

solubility

H2O: 0.1 M, clear, colorless

cation traces

Al: ≤0.0005%
Ca: ≤0.01%
Cu: ≤0.0005%
Fe: ≤0.003%
K: ≤0.005%
Mg: ≤0.003%
NH4+: ≤0.15%
Na: ≤0.02%
Pb: ≤0.005%
Zn: ≤0.0005%

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=CC(=O)NC2=O

InChI

1S/C9H12N2O6/c12-3-4-6(14)7(15)8(17-4)11-2-1-5(13)10-9(11)16/h1-2,4,6-8,12,14-15H,3H2,(H,10,13,16)/t4-,6-,7-,8-/m1/s1

InChI key

DRTQHJPVMGBUCF-XVFCMESISA-N

Gene Information

mouse ... Uck1(22245)

Looking for similar products? Visit Product Comparison Guide

General description

Uridine is a pyrimidine nucleoside which is crucial for the synthesis of RNA and membranes. It helps in normal cell function and growth by forming pyrimidine nucleotide -lipid conjugates.[1]

Application

Uridine has been used to study the functional complementation of nucleoside hydrolase from a protozoan parasite and mammalian uridine phosphorylase on URH1 (uridine hydrolase) of Saccharomyces cerevisiae.[2]
It has been used to check the effect of chronic uridine application on liver metabolism in mice.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chronic Uridine Administration Induces Fatty Liver and Pre-Diabetic Conditions in Mice.
Urasaki Y, et al.
PLoS ONE, 11, e0146994-e0146994 (2016)
Saccharomyces cerevisiae URH1 (encoding uridine-cytidine N-ribohydrolase): functional complementation by a nucleoside hydrolase from a protozoan parasite and by a mammalian uridine phosphorylase.
Mitterbauer R, et al.
Applied and Environmental Microbiology, 68, 1336-1343 (2002)
Purinergic and Pyrimidinergic Signalling: Molecular, Nervous and Urogenitary System Function
Handbook of Experimental Pharmacology (2013)
Qi Qiu et al.
Nature methods, 17(10), 991-1001 (2020-09-02)
Single-cell RNA sequencing offers snapshots of whole transcriptomes but obscures the temporal RNA dynamics. Here we present single-cell metabolically labeled new RNA tagging sequencing (scNT-seq), a method for massively parallel analysis of newly transcribed and pre-existing mRNAs from the same
Ian E Crandall et al.
Journal of medicinal chemistry, 56(6), 2348-2358 (2013-02-16)
Resistance by Plasmodium falciparum to almost all clinically used antimalarial drugs requires the development of new classes of antimalarials. 6-Iodouridine (15), a novel and potent inhibitor of orotidine 5'-monophosphate decarboxylase (ODCase), exhibited efficacy in a mouse model infected by P.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service