Skip to Content
Merck

Skip To

U6000

Uridine 5′-monophosphomorpholidate 4-morpholine-N,N′-dicyclohexylcarboxamidine salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Check Cart for Availability
€ 331,00
5 g
Check Cart for Availability
€ 1.410,00

About This Item

Empirical Formula (Hill Notation):
C13H19N3O9P · C17H32N3O
CAS Number:
Molecular Weight:
686.73
NACRES:
NA.26
PubChem Substance ID:
UNSPSC Code:
12352204
MDL number:
Assay:
≥97% (HPLC)
Biological source:
synthetic (organic)
Form:
powder
Storage temp.:
−20°C

€ 331,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

Quality Segment

assay

≥97% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

C1CCC(CC1)N\C(=N\C2CCCCC2)N3CCOCC3.O[C@H]4[C@@H](O)[C@@H](O[C@@H]4COP(O)(=O)N5CCOCC5)N6C=CC(=O)NC6=O

InChI

1S/C17H31N3O.C13H20N3O9P/c1-3-7-15(8-4-1)18-17(20-11-13-21-14-12-20)19-16-9-5-2-6-10-16;17-9-1-2-16(13(20)14-9)12-11(19)10(18)8(25-12)7-24-26(21,22)15-3-5-23-6-4-15/h15-16H,1-14H2,(H,18,19);1-2,8,10-12,18-19H,3-7H2,(H,21,22)(H,14,17,20)/t;8-,10-,11-,12-/m.1/s1

InChI key

RUAYLFCHPJYFJL-HVVPPSBSSA-N

Application

Uridine 5′-monophosphomorpholidate 4-morpholine-N,N′-dicyclohexylcarboxamidine salt has been used in the synthesis of UDP-TEMPO (uridine 5′-diphospho-4-O-2,2,6,6-tetramethylpiperidine 1-oxyl).

Biochem/physiol Actions

Uridine5′-monophosphomorpholidate 4-morpholine-N,N′-dicyclohexylcarboxamidine salt, also known as UMP-morpholidate, is used as a reactant in the synthesis of uridine5′-(2,3,4-tri-O-acetyl-6-S-acetyl-6-thio-α-d-galactopyranosyldiphosphate). UMP-morpholidate serves as a key reactant in the Khorana-Moffatt procedure for synthesizing uridine diphosphate (UDP)-sugar nucleotides.Uridine 5′-monophosphomorpholidate 4-morpholine-N,N′-dicyclohexylcarboxamidine salt can be used for the synthesis of UDP-azido-GalNAc (uridine diphosphate N-azido acetylgalactosamine).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A1127G7627711233
biological source

synthetic (organic)

biological source

natural (organic)

biological source

synthetic (organic)

biological source

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

assay

≥97% (HPLC)

assay

≥93% (HPLC)

assay

≥90%

assay

97%

form

powder

form

powder

form

powder

form

solid

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters