Skip to Content
Merck
All Photos(1)

Key Documents

U5258

Sigma-Aldrich

Anti-Ubiquitin C-terminal Hydrolase L1 (RA-15) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-UCH-L1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 27 kDa

species reactivity

human, mouse, rat

technique(s)

microarray: suitable
western blot: 1:1,000-1:2,000 using cytosolic fraction (S1) of mouse brain or whole cell extract of human lung carcinoma A549 cell line
western blot: 1:5,000-1:10,000 using cytosolic fraction (S1) of rat brain

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UCHL1(7345)
mouse ... Uchl1(22223)
rat ... Uchl1(29545)

General description

Ubiquitin C-terminal hydrolase L1 (UCH-L1) is encoded by the gene mapped to human chromosome 4p13. It is a 223 amino acid deubiquitinating enzyme, expressed highly in neurons.

Immunogen

synthetic peptide corresponding to amino acids 202-216 located near the C-terminus of rat UCH-L1, conjugated to KLH. This sequence is identical in human, mouse, bovine, porcine, and guinea pig UCH-L1. No homology is found with other known UCH-L isoforms.

Application

Anti-Ubiquitin C-terminal Hydrolase L1 (RA-15) antibody produced in rabbit has been used in immunoblotting.

Biochem/physiol Actions

Ubiquitin C-terminal hydrolase L1 (UCH-L1) hydrolyze C-terminal ubiquityl esters and amides in vitro, which is an essential reaction during cytoplasmic protein degradation. peptide-ubiquityl amides are the most favorable substrates. UCH-L1 also acts as a ubiquitin (Ub) ligase. Mutation in the gene is associated with the development of neurodegenerative diseases, such as Parkinson′s disease, spinocerebellar ataxia (SCA) and Huntington′s disease.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The UCH-L1 gene encodes two opposing enzymatic activities that affect alpha-synuclein degradation and Parkinson's disease susceptibility
Liu Y, et al.
Cell, 111(2), 209-218 (2002)
Reduced expression of the G209A alpha-synuclein allele in familial parkinsonism
Markopoulou K, et al.
Annals of Neurology, 46(3), 374-381 (1999)
Krishnan Sriram et al.
Toxicology and applied pharmacology, 449, 116137-116137 (2022-06-25)
Workers in the oil and gas industry are at risk for exposure to a number of physical and chemical hazards at the workplace. Chemical hazard risks include inhalation of crude oil or its volatile components. While several studies have investigated
Karnam Shruthi et al.
Journal of cellular biochemistry, 120(4), 5962-5973 (2018-10-15)
The ubiquitin-proteasome system (UPS) has been implicated in the pathogenesis of many neurodegenerative diseases. Endoplasmic reticulum (ER) stress is shown to play a pathological role in the development of diabetes and its complications. Hence, the current study is aimed to
Robin K Meray et al.
The Journal of biological chemistry, 282(14), 10567-10575 (2007-01-30)
Deubiquitinating enzymes (DUBs) are negative regulators of protein ubiquitination and play an important role in ubiquitin-dependent processes. Recent studies have found that diverse cellular mechanisms are employed to control the activity of DUBs. Ubiquitin C-terminal hydrolase-L1 (UCH-L1) is a highly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service