Skip to Content
Merck
All Photos(1)

Key Documents

U1753

Sigma-Aldrich

Hydrogen peroxide–Urea adduct

≥97%

Synonym(s):

Carbamide Per hydrate, Carbamide peroxide, Percarbamide, Urea hydrogen peroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CO(NH2)2 · H2O2
CAS Number:
Molecular Weight:
94.07
Beilstein:
3680414
EC Number:
MDL number:
UNSPSC Code:
12352204

vapor pressure

23.3 mmHg ( 30 °C)

Assay

≥97%

composition

H2O2, ~35%

mp

90-93 °C (lit.)

storage temp.

2-8°C

SMILES string

OO.NC(N)=O

InChI

1S/CH4N2O.H2O2/c2-1(3)4;1-2/h(H4,2,3,4);1-2H

InChI key

AQLJVWUFPCUVLO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

A relatively stable source of hydrogen peroxide for preparation of solutions.

Other Notes

An addition compound of urea and hydrogen peroxide.

replaced by

Product No.
Description
Pricing

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 3 - Skin Irrit. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E J Navimipour et al.
Operative dentistry, 38(2), 218-225 (2012-08-04)
This study aimed to evaluate the effect of toothbrushing on enamel surface roughness at three different intervals after daily bleaching treatment. Eighty enamel slabs were initially evaluated for surface roughness and then randomly divided into four groups. The bleaching procedure
Renata Plá Rizzolo Bueno et al.
Journal of applied oral science : revista FOB, 20(3), 335-339 (2012-08-04)
This in vitro study evaluated the influence of two 10% carbamide peroxide bleaching agents - a commercial product (Opalescence PF; Ultradent Products, Inc.) and a bleaching agent prepared in a compounding pharmacy - on the chemical degradation of a light-activated
L E Tam et al.
Operative dentistry, 38(2), 142-150 (2012-09-01)
Although damage to the structural integrity of the tooth is not usually considered a significant problem associated with tooth bleaching, there have been some reported negative effects of bleaching on dental hard tissues in vitro. More studies are needed to
Lidia Yileng Tay et al.
American journal of dentistry, 25(4), 199-204 (2012-10-23)
This parallel, double-blind randomized clinical trial evaluated the 2-year bleaching efficacy and sensitivity produced by at-home (AH) and in-office (IO) bleaching therapies. 60 participants with tooth color darker than C2, without restorations in the anterior dentition and older than 18
Karen Pintado-Palomino et al.
Journal of dentistry, 43(9), 1099-1105 (2015-07-15)
To evaluate the efficacy of experimental proposals of desensitizing agents during tooth bleaching. 140 participants without tooth sensitivity (TS) received 16% carbamide peroxide (14 days-04 h each) (T1) or 35% hydrogen peroxide (single session-45 min) (T2). Participants used concomitantly (10

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service