Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
800 each | Check Cart for Availability | € 1.240,00 |
About This Item
NACRES:
NB.15
UNSPSC Code:
41161502
Manufacturer/tradename:
Nunc 236366
Sterility:
non-sterile
Packaging:
case of 800 ea
€ 1.240,00
Check Cart for Availability
sterility
non-sterile
packaging
case of 800 ea
manufacturer/tradename
Nunc 236366
General description
Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific Nunc Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.
Features and Benefits
- Adhesive seals effectively seal all microplate formats.
- Pressure sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure.
- Working temperature -40°C to +120°C
- Sterile.
- Not autoclavable.
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
1 of 1
This Item | |||
|---|---|---|---|
| description polyester acrylate, non-sterile, case of 800 ea | description rayon acrylate, sterile, case of 50 ea | description rayon acrylate, non-sterile, case of 50 ea | description polyester, non-sterile, case of 100 ea |
| sterility non-sterile | sterility sterile | sterility non-sterile | sterility non-sterile |
| manufacturer/tradename Nunc 236366 | manufacturer/tradename Nunc 241205 | manufacturer/tradename Nunc 249720 | manufacturer/tradename Nunc 236370 |
| packaging case of 800 ea | packaging case of 50 ea | packaging case of 50 ea | packaging case of 100 ea |
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



