Key Documents
TMO174888
Nunclon® cell culture dishes
diam. 60 mm, with lid, Nunclon® Delta treated, case of 120 ea
Synonym(s):
cell culture dish, cell culture treateddish
Select a Size
500 EACH
€1,600.00
€1,600.00
Check Cart for Availability
Select a Size
Change View
500 EACH
€1,600.00
About This Item
UNSPSC Code:
41121812
NACRES:
NB.13
€1,600.00
Check Cart for Availability
material
polystyrene dishes with lids
sterility
sterile; γ-irradiated
packaging
case of 120 ea
manufacturer/tradename
Nunc 174888
diam.
60 mm
General description
  • Large range available, treated for cell culture but with many other uses
  • Optically clear and very flat for use in microscope
  • Non-toxic
  • Certified surface treatment for optimal cell attachment and growth
Legal Information
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)