This product has a working volume of 1.5 mL per well and a culture area of 3.14 cm2. The membrane has a pore density of <0.85 x 108 pores/cm2.
TMO140640
Nunc® cell culture inserts in 6-well Nunclon(TM) Delta surface plate
cell culture treated, polycarbonate, sterile, pore size 0.4 μm, case of 24 ea (supplied in 4 x 6 well plates)
Synonym(s):
multidish membrane insert, treated membrane insert
Select a Size
€278.00
Select a Size
About This Item
€278.00
Recommended Products
material
polycarbonate
sterility
sterile
packaging
case of 24 ea (supplied in 4 x 6 well plates)
manufacturer/tradename
Nunc 140640
pore size
0.4 μm pore size
binding type
cell culture treated
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
-
ich habe 6-wells plate for transwell assay bestellt. aber ich weiss nicht wie viel medium sollte ich in einen Well benutzen, um alle Zellen zu erreichen. Gibt es einen Protocol?
1 answer-
Helpful?
-