T9533
Transforming Growth Factor-α from rat
powder, synthetic, γ-irradiated, suitable for cell culture
Synonym(s):
TGF-α
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C244H367N71O71S6
CAS Number:
Molecular Weight:
5623.35
MDL number:
UNSPSC Code:
12352200
biological source
synthetic
sterility
γ-irradiated
Assay
~90% (HPLC)
form
powder
potency
0.02-2.0 ng/mL EC50 (using Balb/MK 1)
mol wt
5.5 kDa
composition
Peptide content, ~80%
storage condition
avoid repeated freeze/thaw cycles
technique(s)
cell culture | mammalian: suitable
impurities
endotoxin, tested
UniProt accession no.
storage temp.
−20°C
Gene Information
rat ... Tgfa(24827) , Tgfa(24827)
Looking for similar products? Visit Product Comparison Guide
Amino Acid Sequence
Val-Val-Ser-His-Phe-Asn-Lys-Cys-Pro-Asp-Ser-His-Thr-Gln-Tyr-Cys-Phe-His-Gly-Thr-Cys-Arg-Phe-Leu-Val-Gln-Glu-Glu-Lys-Pro-Ala-Cys-Val-Cys-His-Ser-Gly-Tyr-Val-Gly-Val-Arg-Cys-Glu-His-Ala-Asp-Leu-Leu-Ala [Disulfide Bridges: 8-21; 16-32; 34-43]
Biochem/physiol Actions
Transforming growth factor-α (TGF-α), is a 5.5 kDa polypeptide containing 50 amino acids in its mature form. It was originally identified as an agent that reversibly confers a transformed phenotype upon normal non-neoplastic cells, such as normal rat kidney fibroblasts. This activity requires the presence of transforming growth factor-β (TGF-β), which potentiates the action of TGF-α via a separate receptor. TGF-α is synthesized by monocytes, keratinocytes and many tissues and tumors. Mice genetically engineered to be lacking TGF-α expression showed no abnormalities except for the detection of a "wavy" hair coat. TGF-α exerts its action through the EGF receptor.
comparable product
Product No.
Description
Pricing
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service