Skip to Content
Merck
All Photos(1)

Key Documents

T9375

Sigma-Aldrich

Thymidine 5′-diphosphate sodium salt

≥96% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€115.00
25 MG
€417.00
50 MG
€698.00

€115.00


Estimated to ship on28 May 2025



Select a Size

Change View
5 MG
€115.00
25 MG
€417.00
50 MG
€698.00

About This Item

Empirical Formula (Hill Notation):
C10H16N2O11P2 · xNa+
CAS Number:
Molecular Weight:
402.19 (free acid basis)
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

€115.00


Estimated to ship on28 May 2025


biological source

synthetic (organic)

Assay

≥96% (HPLC)

form

powder

solubility

water: 50 mg/mL, clear to very slightly hazy, colorless

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].CC1=CN([C@H]2C[C@H](O)[C@@H](COP([O-])(=O)OP([O-])([O-])=O)O2)C(=O)NC1=O

InChI

1S/C10H16N2O11P2.3Na/c1-5-3-12(10(15)11-9(5)14)8-2-6(13)7(22-8)4-21-25(19,20)23-24(16,17)18;;;/h3,6-8,13H,2,4H2,1H3,(H,19,20)(H,11,14,15)(H2,16,17,18);;;/q;3*+1/p-3/t6-,7+,8+;;;/m0.../s1

InChI key

HBYPEEZBYHQLDM-SPSULGLQSA-K

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Thymidine5′-diphosphate sodium salt has been used as a substrate to investigate the efficacy of an online methodology for direct monitoring of nucleoside monophosphate and nucleotide diphosphate phosphorylation.[1]

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service