Skip to Content
Merck
All Photos(1)

Key Documents

T8641

Sigma-Aldrich

Glyceryl tridodecanoate

~98%

Synonym(s):

1,2,3-Tridodecanoylglycerol, 1,2,3-Trilauroylglycerol, Glycerol trilaurate, Glyceryl trilaurate, Tridodecanoin, Trilaurin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)10COOCH2]2CHOCO(CH2)10CH3
CAS Number:
Molecular Weight:
639.00
Beilstein:
1730452
EC Number:
MDL number:
UNSPSC Code:
12352211

Assay

~98%

mp

46.5 °C (lit.)

storage temp.

−20°C

SMILES string

CCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCC)OC(=O)CCCCCCCCCCC

InChI

1S/C39H74O6/c1-4-7-10-13-16-19-22-25-28-31-37(40)43-34-36(45-39(42)33-30-27-24-21-18-15-12-9-6-3)35-44-38(41)32-29-26-23-20-17-14-11-8-5-2/h36H,4-35H2,1-3H3

InChI key

VMPHSYLJUKZBJJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

David A Pink et al.
The Journal of chemical physics, 132(5), 054502-054502 (2010-02-09)
We investigated theoretically two competing published scenarios for the melting transition of the triglyceride trilaurin (TL): those of (1) Corkery et al. [Langmuir 23, 7241 (2007)], in which the average state of each TL molecule in the liquid phase is
A Svensson et al.
International journal of pharmaceutics, 281(1-2), 107-118 (2004-08-04)
The incorporation of drugs into Gelucires has been reported to increase the dissolution rate of poorly soluble drugs, often leading to improved drug bioavailability. In pharmaceutical applications, it is important to know how the excipient interacts with the drug, and
K Takahashi et al.
Enzyme, 32(4), 235-240 (1984-01-01)
Modified lipoprotein lipase catalyzed the synthesis of trilaurin from mono- or diacylglycerol and fatty acid and also the synthesis of ester from fatty acid and alcohol in benzene. In the ester synthesis reaction, the longer the chain length of fatty
T L Carlson et al.
Biochimica et biophysica acta, 1083(3), 221-229 (1991-06-03)
Age-matched Watanabe (WHHL) and New Zealand White (NZW) rabbits were fed a coconut oil-enriched diet (14%, w/w) for 2 weeks. Lipid and apolipoprotein (apo) A-I levels in plasma and lipoprotein fractions were monitored. Within 3 days after the start of
S P Vyas et al.
The Journal of pharmacy and pharmacology, 58(3), 321-326 (2006-03-16)
In this study we developed emulsomes, a novel lipoidal vesicular system with an internal solid fat core surrounded by a phospholipid bilayer. Plain emulsomal formulations composed of solid lipid (trilaurin or tristearin), cholesterol and soya phosphatidylcholine and stearylamine containing cationic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service