Skip to Content
Merck

T8024

Tetrodotoxin

≥98% (HPLC), powder

Synonym(s):

Fugu poison, Maculotoxin, TTX, Tarichatoxin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H17N3O8
CAS Number:
Molecular Weight:
319.27
EC Number:
224-458-8
UNSPSC Code:
12352200
PubChem Substance ID:
Beilstein/REAXYS Number:
49176
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white

solubility

dilute citrate or acetate buffer: 1 mg/mL (dissolve at pH 4-5. Aqueous solutions, pH 4-5, are stable if stored frozen.), strong acid or alkaline solutions: unstable (destroyed by boiling at pH 2.)

storage temp.

2-8°C

SMILES string

[H][C@]12OC3(O)OC(C4[C@@H](O)NC(=N)NC4([C@@H]1O)[C@@H]3O)[C@@]2(O)CO

InChI

1S/C11H17N3O8/c12-8-13-6(17)2-4-9(19,1-15)5-3(16)10(2,14-8)7(18)11(20,21-4)22-5/h2-7,15-20H,1H2,(H3,12,13,14)/t2?,3-,4?,5+,6-,7+,9+,10?,11?/m1/s1

InChI key

CFMYXEVWODSLAX-IXUBDLNJSA-N

Gene Information

rat ... Scnn1g(24768)

Application

Tetrodotoxin has been used as standard during HPLC-based analysis of biotoxins from P. punctata tentacles whole crude extract (PHY-N). Tetrodotoxin has also been used to study its effects on low-threshold transient (LTT) Ca2+ currents in rat hippocampal interneurons.

Biochem/physiol Actions

Reversible, selective blocker of Na+ channels.
Reversible, selective blocker of Na+ channels; blocks propagation of impulses in excitable membranes. Used to characterize sodium channels in excitable membranes and to study the role of sodium channels in normal physiology and disease.

Preparation Note

Tetrodotoxin is soluble in dilute citrate or acetate buffer at 1 mg/ml (pH 4-5). Aqueous solutions are stable at pH 4-5 if stored frozen. However, tetrodotoxin is unstable in strong acid or alkaline solutions and is destroyed by boiling at pH2.


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 1 Oral

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Simone Berkel et al.
Human molecular genetics, 21(2), 344-357 (2011-10-14)
Mutations in the postsynaptic scaffolding gene SHANK2 have recently been identified in individuals with autism spectrum disorder (ASD) and intellectual disability. However, the cellular and physiological consequences of these mutations in neurons remain unknown. We have analyzed the functional impact
D D Fraser et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 11(9), 2812-2820 (1991-09-01)
Interneurons from the CA1 lacunosum-moleculare (L-M) region were isolated by trypsin-hyaluronidase treatment and mechanical trituration of the L-M. Interneurons isolated in this manner were multipolar with several dendritic processes and could be distinguished from CA1 pyramidal neurons. The properties of
Rocky Chau et al.
Aquatic toxicology (Amsterdam, Netherlands), 104(1-2), 61-72 (2011-05-06)
The potent neurotoxin tetrodotoxin (TTX) has been identified from taxonomically diverse marine organisms. TTX possesses a unique cage-like structure, however, its biosynthesis has yet to be elucidated. Biosynthetic studies in the TTX-producing newt Taricha torosa, and in bacterial genera, including