Skip to Content
Merck
All Photos(1)

Key Documents

T7927

Sigma-Aldrich

L-Tyrosine Decarboxylase Apoenzyme from Streptococcus faecalis

<0.005 unit/mg solid (without pyridoxal 5-phosphate), ≥0.05 unit/mg solid (with excess pyridoxal 5-phosphate)

Synonym(s):

L-(−)-Tyrosine Apodecarboxylase, L-Tyrosine Carboxy-lase apoenzyme, TAD

Sign Into View Organizational & Contract Pricing

Select a Size

25 UNITS
€277.00
50 UNITS
€480.00
250 UNITS
€1,720.00

€277.00


Estimated to ship on30 April 2025



Select a Size

Change View
25 UNITS
€277.00
50 UNITS
€480.00
250 UNITS
€1,720.00

About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

€277.00


Estimated to ship on30 April 2025


biological source

Streptococcus faecalis

Quality Level

form

powder

specific activity

≥0.05 unit/mg solid (with excess pyridoxal 5-phosphate)
<0.005 unit/mg solid (without pyridoxal 5-phosphate)

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

L-Tyrosine decarboxylase apoenzyme from Streptococcus faecalis has been used:
  • in a study to purify and characterize tyrosine decarboxylase and aromatic-L-amino-acid decarboxylase[1]
  • in a study to investigate the stereospecificity of sodium borohydride reduction of tyrosine decarboxylase[2]
  • in protein synthesis studies[3]

Biochem/physiol Actions

L-Tyrosine Decarboxylase Apoenzyme is an aromatic L-amino acid decarboxylase.[4] The enzyme activity of tyrosine decarboxylase (TDC) is dependent on the cofactor pyridoxal 5-phosphate (PLP).[5] It catalyzes the decarboxylation of dihydroxyphenylalanine.[4]

Other Notes

Dried cells grown on vitamin B6-deficient medium.

Unit Definition

One unit will liberate 1.0 μmole of CO2 from L-tyrosine per min at pH 5.5 at 37 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haixia Zhu et al.
Scientific reports, 6, 27779-27779 (2016-06-14)
Tyrosine decarboxylase (TDC) is a pyridoxal 5-phosphate (PLP)-dependent enzyme and is mainly responsible for the synthesis of tyramine, an important biogenic amine. In this study, the crystal structures of the apo and holo forms of Lactobacillus brevis TDC (LbTDC) were
Gunsalus, I.C., and Smith, R.A.
Methods in Enzymology, 3, 963-963 (1957)
T Miura et al.
International journal of cancer, 46(5), 931-934 (1990-11-15)
Phenolic melanin precursors can be utilized for the development of anti-melanoma agents. The sulphur homologue of tyrosine, 4-S-cysteinylphenol (CP) and its decarboxylation product, 4-S-cysteaminylphenol (CAP) were shown to be substrates of melanoma tyrosinase, forming melanin-like pigment. Both, but in particular
J C Vederas et al.
The Journal of biological chemistry, 254(12), 5053-5057 (1979-06-25)
Sodium boro[3H]hydride reduction of tyrosine decarboxylase from Streptococcus faecalis followed by complete hydrolysis of the enzyme produces epsilon-[3H]pyridoxyllysine. Degradation of this material to [4'-3H]pyridoxamine and stereochemical analysis with apoaspartate aminotransferase shows that the re side at C-4' of the cofactor
Impaired growth and force production in skeletal muscles of young partially pancreatectomized rats: a model of adolescent type 1 diabetic myopathy?
Gordon CS, et al.
PLoS ONE (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service