Skip to Content
Merck

Skip To

T7201

Thrombin from bovine plasma

lyophilized powder, ≥2,000 NIH units/mg protein (E1%/280 = 19.5)

Synonym(s):

Factor IIa

Sign In to View Organizational & Contract Pricing.

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
MDL number:
Specific activity:
≥2,000 NIH units/mg protein (E1%/280 = 19.5)


form

lyophilized powder

specific activity

≥2,000 NIH units/mg protein (E1%/280 = 19.5)

mol wt

heavy chain ~31 kDa, light chain ~6 kDa

storage temp.

−20°C

General description

Thrombin is the final coagulation protease in regard to hemostasis, promoting both procoagulant and anticoagulant effects. Thrombin consists of A-chain and a catalytic B-chain with the catalytic pocket comprising of His57, Asp102, Ser195 and Ser214. It has a fibrinogen recognition site (FRS) and a heparin binding site (HBS) in the C-terminus. The main function of thrombin is cleavage of fibrinogen to fibrin. It coordinates the coagulation cascade and binds to diverse substrates. It coordinates with protease-activated receptors (PARs) to accomplish the physiological functions. High levels of thrombin elicit neurotoxicity in dopaminergic neurons and contribute to the progression of Parkinson′s disease. Altered thrombin levels modulates coagulation pathway in multiple sclerosis. Patients with coronary artery disease (CAD) show elevated levels of thrombin.

Application

Thrombin is used for site specific cleavage of recombinant fusion proteins containing an accessible thrombin recognition site for removal of affinity tags. Thrombin has been used in a study to assess a novel antiplatelet therapy in acute coronary syndromes. [1]
Thrombin from bovine plasma has been used in the cleavage of the maltose binding protein tag in mini-calpain-5 fusion protein and in thrombin/hirudin spiking experiments in serum samples.

Biochem/physiol Actions

Serine protease that selectively cleaves Arg-Gly bonds in fibrinogen to form fibrin and fibrinopeptides A and B.

Physical form

Lyophilized from saline sodium citrate buffer, pH 6.5

Preparation Note

Traditional thrombin products are activated with bovine brain, whereas T7201 is activated with bovine lung and does not contain any bovine brain products.

Analysis Note

Activity is expressed in NIH units obtained by direct comparison to a NIH Thrombin Reference Standard, Lot K.
The NIH assay procedure uses 0.2 mL of diluted plasma (1:1 with saline) as a substrate and 0.1 mL of thrombin sample (stabilized in a 1% buffered albumin solution) based on a modification of the method of Biggs. Only clotting times in the range of 15-25 seconds are used for determining thrombin concentrations.

Other Notes

Activity is expressed in NIH units obtained by direct comparison to a NIH thrombin reference standard.
View more information on thrombin at www.sigma-aldrich.com/enzymeexplorer.


Still not finding the right product?

Explore all of our products under Thrombin from bovine plasma


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters