Merck
Skip To
T6948
Sartorius pipette tips
volume range 100-5000 μL, standard, bulk packed, non-sterile
Synonym(s):
Pipette Tips, Sartorius Pipette Tips, Sartorius Tips
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
1 pkg
Check Cart for Availability
€ 18,30
About This Item
NACRES:
NB.01
UNSPSC Code:
41121600
Volume:
100—5000 μL
Sterility:
non-sterile
Feature:
DNase free, RNase free, barrier: no, endotoxin free
Packaging:
pack of 100 ea (Color coded tip trays)
Manufacturer/tradename:
Sartorius 780300
€ 18,30
Check Cart for Availability
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
colorless
sterility
non-sterile
feature
DNase free, RNase free, barrier: no, endotoxin free
packaging
pack of 100 ea (Color coded tip trays)
manufacturer/tradename
Sartorius 780300
maximum volume
5 mL
volume range
100-5000 μL
volume
100—5000 μL
General description
The non-filtered Optifit Tips are designed for best possible fitting and sealing to secure the highest possible accuracy and precision. These pipette tips enable ergonomic and light tip attachment.
Optifit Tips are available in various packaging types for all your needs:
  • Single Tray racks – purity certified tip racks
  • Refill Packs - purity certified and ecological refill packages
  • Refill Towers - space-saving and ecological refill packages
  • Flexibulk boxes - space-saving, ecological and economical bulk boxes
  • Standard bulk – economical bulk boxes
While Optifit Tip trays are colour coded for easy matching with a corresponding, colour coded Sartorius pipette, they do also match with most other pipette due to their universal design.
Strict quality standards, ISO 9001 and ISO 13485, are followed from tip R&D to production and delivery. The purity certified tips are manufactured in ISO 8 classified clean room conditions to avoid any contaminations and are tested for DNase, RNase and endotoxins per each tip lot. Pre-sterilised tips are also available for the purest laboratory work. Sartorius offers a selection of pipette tips, extended and large volume tips, for special needs.
Other Notes
Browse all pipette tips
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
Z741651Z757721Z741648
sterility
non-sterile
sterility
non-sterile
sterility
sterile
sterility
non-sterile
volume
100—5000 μL
volume
500—10000 μL
volume
5—350 μL
volume
100—5000 μL
manufacturer/tradename
Sartorius 780300
manufacturer/tradename
Sartorius 780310
manufacturer/tradename
Sartorius 790353
manufacturer/tradename
Sartorius 780304
feature
DNase free, barrier: no, endotoxin free, RNase free
feature
DNase free, RNase free, barrier: no, endotoxin free
feature
DNase free, RNase free, barrier: no, endotoxin free
feature
DNase free, RNase free, barrier: no, endotoxin free
material
colorless
material
colorless , colorless tip
material
colorless
material
colorless
packaging
pack of 100 ea (Color coded tip trays)
packaging
pack of 100 ea
packaging
pack of 96 ea (Color coded tip trays)
packaging
pack of 50 ea
Still not finding the right product?
Explore all of our products under Sartorius pipette tips
Choose from one of the most recent versions:
Certificates of Analysis (COA)