Unfortunately, an Xbal restriction site is not present. Please see the link below to review the product datasheet, which includes information on restriction sites available for this item.
https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/284/891/t6701bul.pdf
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
2 μg | Check Cart for Availability | € 195,00 |
About This Item
NACRES:
NA.51
UNSPSC Code:
41106609
General description
The TargeTron Vector pNL9164 is a 9,295 bp expression vector for use with the TargeTron Gene Knockout System (product number TA0100). This circularized vector can be used for targeted gene knockouts in gram-positive organisms such as Staphyloccus aureus. As supplied, the pNL9164 vector is re-targeted to the S. aureus hsa gene. The hsa gene encodes a member of the HU family of bacterial histone-like proteins. For validation in S. aureus, this vector can be used directly to knockout hsa without any further modifications.
Legal Information
TargeTron is a trademark of InGex, LLC
1 of 1
This Item | |||
|---|---|---|---|
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −70°C |
| shipped in wet ice | shipped in - | shipped in wet ice | shipped in - |
| concentration 100 ng/μL | concentration - | concentration - | concentration - |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 100 |
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Hi, does pNL9164 have a XbaI restriction site located within the group II intron RNA coding sequence?
1 answer-
Helpful?
-



