Skip to Content
Merck
All Photos(1)

Key Documents

T6638

Sigma-Aldrich

Tolnaftate

Synonym(s):

Methyl-(3-methylphenyl)carbamothioic acid O-2-naphthyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17NOS
CAS Number:
Molecular Weight:
307.41
EC Number:
MDL number:
UNSPSC Code:
51302004
PubChem Substance ID:
NACRES:
NA.85

form

powder

storage condition

(Keep container tightly closed in a dry and well-ventilated place.)

color

white to off-white

solubility

chloroform: soluble 50 mg/mL

antibiotic activity spectrum

fungi

Mode of action

enzyme | inhibits

storage temp.

2-8°C

SMILES string

CN(C(=S)Oc1ccc2ccccc2c1)c3cccc(C)c3

InChI

1S/C19H17NOS/c1-14-6-5-9-17(12-14)20(2)19(22)21-18-11-10-15-7-3-4-8-16(15)13-18/h3-13H,1-2H3

InChI key

FUSNMLFNXJSCDI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tolnaftate is used to study the mechanism of ergosterol (sterol) biosynthesis at the rate-limiting step of squalene epoxidase inhibition. Tolnaftate is clinically used to treat cutaneous infections such as athlete′s foot, jock itch, and ringworm. It also has been used as a comparator compound for antifungal efficiency studies.

Biochem/physiol Actions

Tolnaftate prevents ergosterol biosynthesis by inhibiting squalene epoxidase. It has also been reported to distort the hyphae and to stunt mycelial growth in susceptible organisms. Tolnaftate inhibits biosynthesis of aflatoxin.

Packaging

1G,5G

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Kobayashi et al.
Chemical & pharmaceutical bulletin, 46(11), 1797-1802 (1998-12-10)
The enhancing effects of various vehicles on the in vitro permeation of a hydrophilic model drug, 5-fluorouracil (5-FU), or a lipophilic model drug, tolnaftate (TN), through human nail plates were investigated using a modified side-by-side diffusion cell. Tip pieces from
A K Dash et al.
Journal of pharmaceutical sciences, 91(4), 983-990 (2002-04-12)
Ethylcellulose microspheres containing tolnaftate (I) were prepared by the emulsion-solvent evaporation technique. An X-ray powder diffractometric method was developed to quantify the content of crystalline I in these microspheres. X-ray lines of I with d-spacings of 5.5 and 4.2 A
Toma Kezutyte et al.
Acta poloniae pharmaceutica, 67(4), 327-334 (2010-07-20)
Abstract: Tolnaftate, an antifungal of thiocarbamate class, is used topically in 1% formulations. Its penetration into skin layers is a prerequisite for tolnaftate action against dermatophytes. The aim of this work was to optimize and validate a simple, rapid, accurate
T Ohmi et al.
Arzneimittel-Forschung, 41(8), 847-851 (1991-08-01)
In vitro and in vivo antifungal activities of the new imidazole derivative latoconazole ((+-)-(E)-[4-(2-chlorophenyl)-1,3-dithiolan-2-ylidene]-1- imidazolylacetonitrile, NND-318; CAS 101530-10-3) were studied in comparison with three major topical agents, clotrimazole, bifonazole and tolnaftate. The in vitro activity of latoconazole against dermatophytes was
Toma Kezutyte et al.
Acta poloniae pharmaceutica, 68(6), 965-973 (2011-12-01)
Five fatty acids (oleic, linoleic, myristic, lauric and capric) were incorporated in 10% (w/w) into ointment formulation and their influence on lipophilic model drug tolnaftate release in vitro and enhancing effect on tolnaftate penetration into epidermis and dermis of human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service